Sigma-Aldrich
Analytical / Chromatography > Titration > HYDRANAL® Karl Fischer Reagents > KF Oven Reagents
  • Pricing & availability is not currently available.
    Questions? Contact Technical Service
  • USA Home
  • Product Directory
  • Analytical/Chromatography
  • Analytical Reagents
  • Titration
  • Karl Fischer Titration Reagents (HYDRANAL)
  • KF Reagents for Special Applications
  • Special Reagents for Karl Fischer Oven
Special Reagents for Karl Fischer Oven
Analytical Reagents Search
Product #
Description
Add to Cart
34739 HYDRANAL®-Coulomat AG-Oven reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm
  • pricing
  • 34840 HYDRANAL®-Coulomat CG reagent for coulometric KF titration (catholyte solution)
  • pricing
  • 34788 HYDRANAL®-Humidity absorber Drying agent for air and gases for KF titration (with indicator)
  • pricing
  • 34241 HYDRANAL®-Molecular sieve 0.3 nm Drying agent for air and gases for KF titration
  • pricing
  • 34693 HYDRANAL®-Water Standard KF-Oven, 140-160°C control of KF oven (solid standard, water content ~5%, exact value on CoA)
  • pricing
  • 34748 HYDRANAL®-Water Standard KF-Oven 220°C-230°C control of KF oven (solid standard, water content 5.55 ± 0.05%, exact value on CoA)
  • pricing
  • your favorite gene search