Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
HYDRANAL®
HYDRANAL Manual Request
HYDRANALBOOK
HYDRANAL® is the gold standard reagent line for pyridine-free moisture determination by Karl-Fischer titration.
The HYDRANAL®-Manual is your one-stop reference for everything you need to know about HYDRANAL®
As always, the HYDRANAL®-Manual laboratory hand-book provides practical, up-to-date information on Karl-Fischer titration including:
  • Principles of Karl Fischer titration• Standard procedures for volumetric and coulometric titration
  • Selection of the appropriate solvents for the sample
  • Sample handling techniques
  • Complete HYDRANAL® product listing
This year, we've updated the HYDRANAL®-Manual with:
  • New! HYDRANAL® products, like HYDRANAL®-Medium K, a safer solvent for the analysis of Aldehydes and Ketones and HYDRANAL®-Solver (Crude) Oil a solvent for the analysis of Crude Oil samples
  • New! HYDRANAL® Applications
  • New! HYDRANAL® Lab Reports
  • New! HYDRANAL® Literature