Sigma-Aldrich
Analytical / Chromatography > Titration > HYDRANAL® Karl Fischer Reagents > Learning Center > Theory of Karl Fischer Titration
HYDRANAL®
Theory of Karl Fischer Titration
Introduction
Sigma-Aldrich is a pioneer in the supply of safe, reliable and easy-to-use pyridine-free Karl Fischer reagents.
Water can affect product quality, texture, shelf life, chemical stability and reactivity. The Karl Fischer titration is a universally acknowledged method for measuring water content in all types of substances including chemicals, oils, pharmaceuticals and food. In 1979, researcher Dr. Eugen Scholz improved the Karl Fischer titration by replacing noxious pyridine with imidazole.
This innovation became the foundation of HYDRANAL®, the world’s leading pyridine-free reagents for Karl Fischer titration. Since Dr. Scholz’s pioneering research and with a spirit of ongoing product improvement, Sigma-Aldrich has launched a wide range of Karl Fischer reagents for both volumetric and coulometric titrations for nearly all types of samples.
Advantages of HYDRANAL® reagents:
  • High-speed titrations
  • Stable end points
  • Accurate results
  • No unpleasant odor
  • Lower toxicity for increased safety and decreased environmental impact
  • Long shelf life
  • Wide applicability
Table 1: HYDRANAL® Product Line at a Glance
HYDRANAL®-Composite The most frequently used pyridine-free Karl Fischer reagents for one-component volumetric titration
HYDRANAL®-Solvent / Titrant Volumetric titration with two-component reagents for higher accuracy and stability
HYDRANAL®-Coulomat Coulometric titration for samples with low water content
HYDRANAL®-K types Specially designed reagents for determining water in aldehydes and ketones
HYDRANAL®-E types Reduced toxicity over ethanol-containing reagents for both volumetric and coulometric titrations
HYDRANAL®-Water Standards Standards with certified water content for titer determination
In the following chapters, you will find the basic principles and possibilities of Karl Fischer titration.
Additional technical information can be obtained by requesting our HYDRANAL®-Manual, Multimedia Guide CD, Literature or Applications. For personal assistance, please feel free to contact our Karl Fischer helplines.
Chapter 1 Basic principles of Karl Fischer titration
  • HYDRANAL® reagents
  • Standard procedures for Karl Fischer titration
  • Chemical fundamentals behind Karl Fischer titration (working medium, pH value)
Chapter 2 Basic forms of Karl Fischer titration
  • Volumetric titration with one-component reagents
  • Volumetric titration with two-component reagents
  • Coulometric titration
Chapter 3 Variations of Karl Fischer titration
  • Overview of variations of Karl Fischer titration
  • Reagents for aldehydes and ketones
  • Reagents for non-polar substances
  • Reagents for polar substances
  • Reagents for acids and bases
  • Reagents for Karl Fischer Oven
Literature
[1] Eugen Scholz „Karl-Fischer-Titration“
  Springer-Verlag, Berlin, Heidelberg, New York, Tokyo (1984), 140 pages
  ISBN 3-540-12846-8 German Edition
  ISBN 3-540-13734-3 English Edition
Back to Top
Back to HYDRANAL® home
ORDERING
SERVICE & SUPPORT
CORPORATE
    Follow us on Twitter!  Join us on Facebook!  Join us on LinkedIn!  Watch our videos on YouTube!
Copyrights © 2010 Sigma-Aldrich Co. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.