Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Titration
Volumetric Titration with one-component reagents
Advantages of one-component titration:
  • Determination of unlimited range of water content
  • Convenient and easy to use
  • Long shelf life
Product range HYDRANAL® volumetric one-component reagents
Composition
HYDRANAL®-Composite is a one-component Karl Fischer reagent for volumetric Karl Fischer titration. It contains all reactants: iodine, sulfur dioxide and the bases imidazole and 2-methylimidazole, dissolved in diethyleneglycol monoethyl ether (DEGEE). Using 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals, which can interfere with the performance of the titrator. We have applied for a patent for this new formulation.
Stability
HYDRANAL®-Composite is stabilized with DEGEE instead of 2-methoxyethanol. DEGEE improves stability; the concentration loss is less than 5 % per year.
Toxicity
HYDRANAL®-Composites are free of pyridine and 2-methoxyethanol.
Using HYDRANAL®-Composite
We recommend the following procedure for titrations with HYDRANAL®-Composite (Figure 2.1):
  1. Fill the burette with HYDRANAL®-Composite
  2. Add HYDRANAL®-Methanol dry or HYDRANAL®-Methanol Rapid or HYDRANAL®-CompoSolver E to the titration vessel
  3. Titrate to dryness with HYDRANAL®-Composite
  4. Add sample
  5. Titrate the water content with HYDRANAL®-Composite
Volumetric one-component titration with HYDRANAL®-Composite Figure 2.1: Volumetric one-component titration with HYDRANAL®-Composite.
Back to Introduction page
Back to HYDRANAL® Home