Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Karl Fischer Theory
Overview of Variations
The Karl Fischer titration can be applied to a wide variety of substances. The differences in sample properties, especially their solubility, can influence the course of the Karl Fischer reaction. There are a number of ways to adjust the working conditions in order to enhance solubility and therefore enable a direct Karl Fischer titration of a sample. Many of these variations are described in this section.
  • Fats, oils and long-chained hydrocarbons have limited solubility in methanol. Solubility can be improved by the addition of long-chained alcohols or chloroform to the working medium.
  • Polar substances, like proteins, carbohydrates and inorganic salts are either insoluble or dissolve only slightly in organic solvents. Addition of formamide improves solubility or dispersion of these samples.
  • With certain samples, methanol can cause interfering side reactions and should be replaced with other solvents.
  • Acids and bases can alter the pH of the Karl Fischer system outside the desirable range and must be neutralized.
  • Natural materials (soils, grains, etc.) release water slowly. Titration at 50 °C or in boiling methanol accelerates the dissolution of the sample and the subsequent extraction of water.
  • Ovens are used to draw out moisture from solids that only release water at temperatures higher than 60 °C. The evaporated water is carried over into the titration cell by a dried inert gas.
If the working conditions of the titration are modified, the requirements of the Karl Fischer reaction must be considered and stoichiometry must be maintained.
Back to Introduction page
Back to HYDRANAL® Home