Sigma-Aldrich
Analytical / Chromatography > Titration > HYDRANAL® Karl Fischer Reagents > Learning Center > Theory of Karl Fischer Titration > Overview of Variations
Karl Fischer Theory
Overview of Variations
The Karl Fischer titration can be applied to a wide variety of substances. The differences in sample properties, especially their solubility, can influence the course of the Karl Fischer reaction. There are a number of ways to adjust the working conditions in order to enhance solubility and therefore enable a direct Karl Fischer titration of a sample. Many of these variations are described in this section.
  • Fats, oils and long-chained hydrocarbons have limited solubility in methanol. Solubility can be improved by the addition of long-chained alcohols or chloroform to the working medium.
  • Polar substances, like proteins, carbohydrates and inorganic salts are either insoluble or dissolve only slightly in organic solvents. Addition of formamide improves solubility or dispersion of these samples.
  • With certain samples, methanol can cause interfering side reactions and should be replaced with other solvents.
  • Acids and bases can alter the pH of the Karl Fischer system outside the desirable range and must be neutralized.
  • Natural materials (soils, grains, etc.) release water slowly. Titration at 50 °C or in boiling methanol accelerates the dissolution of the sample and the subsequent extraction of water.
  • Ovens are used to draw out moisture from solids that only release water at temperatures higher than 60 °C. The evaporated water is carried over into the titration cell by a dried inert gas.
If the working conditions of the titration are modified, the requirements of the Karl Fischer reaction must be considered and stoichiometry must be maintained.
  • Reagents for aldehydes and ketones
  • Reagents for non-polar substances
  • Reagents for polar substances
  • Reagents for acids and bases
  • Reagents for Karl Fischer Oven
Back to Introduction page
Back to HYDRANAL® Home
ORDERING
SERVICE & SUPPORT
CORPORATE
    Follow us on Twitter!  Join us on Facebook!  Join us on LinkedIn!  Watch our videos on YouTube!
Copyrights © 2010 Sigma-Aldrich Co. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.