Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
HYDRANAL®
One-component Reagents
Ordering Information
Composition
HYDRANAL®-Composite is the world's most frequently used pyridine-free Karl Fischer reagent for one-component titration. It contains all the reactants, i.e. iodine, sulfur dioxide and imidazole, dissolved in diethyleneglycol monoethyl ether (DEGEE).
Our HYDRANAL R & D laboratory has recently developed an improved formulation for HYDRANAL®-Composite by using a mixture of imidazole and 2-methylimidazole instead of imidazole only. Significant improvements in the properties of HYDRANAL®-Composite have been obtained as a result of this change.
Stability
HYDRANAL®-Composite is stabilized with DEGEE. The loss of titre is approx. 5% a year. Other Karl Fischer reagents contain 2-methoxyethanol (2-ME).
Toxicity
All HYDRANAL®-Composites are pyridine free and free of 2-methoxyethanol.
Advantages of the one-component titration
  • Unlimited water capacity.
  • Three different titres available.
  • Also convenient for the analysis of ketones.
  • Widely used: many different applications for the determination of 1 - 100 mg water.
  • Suitable for customary volumetric Karl Fischer titrators.
HYRDRANAL videos
Learn more - watch our
HYDRANAL videos
View Product List and Ordering Information