Sigma-Aldrich
Analytical / Chromatography > Titration > HYDRANAL® Karl Fischer Reagents > One-component Reagents
HYDRANAL®
One-component Reagents
Ordering Information
Composition
HYDRANAL®-Composite is the world's most frequently used pyridine-free Karl Fischer reagent for one-component titration. It contains all the reactants, i.e. iodine, sulfur dioxide and imidazole, dissolved in diethyleneglycol monoethyl ether (DEGEE).
Our HYDRANAL R & D laboratory has recently developed an improved formulation for HYDRANAL®-Composite by using a mixture of imidazole and 2-methylimidazole instead of imidazole only. Significant improvements in the properties of HYDRANAL®-Composite have been obtained as a result of this change.
Stability
HYDRANAL®-Composite is stabilized with DEGEE. The loss of titre is approx. 5% a year. Other Karl Fischer reagents contain 2-methoxyethanol (2-ME).
Toxicity
DEGEE is non-harzardous unlike 2-ME which is fetotoxic and teratogenic. All HYDRANAL®-Composites are pyridine free and free of 2-methoxyethanol. Toxicological studies are done.
Advantages of the one-component titration
  • Unlimited water capacity.
  • Three different titres available.
  • Also convenient for the analysis of ketones.
  • Widely used: many different applications for the determination of 1 - 100 mg water.
  • Suitable for customary volumetric Karl Fischer titrators.
HYRDRANAL videos
Learn more - watch our
HYDRANAL videos
View Product List and Ordering Information
Literature
English
HYDRANAL®-Composite Improved one-component reagent for volumetric Karl Fischer Titration
HYDRANAL-Keto Solver HYDRANAL-Keto Solver ... For Volumetric Moisture detemination
HYDRANAL®-Methanol Rapid For fast Karl Fischer titrations
HYDRANAL®-Medium K Excellent performance without toxic components
Deutsch
HYDRANAL®-Composite Verbessertes Einkomponentenreagenz für die volumetrische
Karl-Fischer-Titration
HYDRANAL®-Methanol Rapid Für reaktionsschnelle Karl Fischer-Titration
HYDRANAL®-KetoSolver Für die volumetrische Wasserbestimmung
HYDRANAL®-Medium K  Exzellente Performance ohne toxische Komponenten