Sigma-Aldrich
Analytical / Chromatography > Titration > HYDRANAL® Karl Fischer Reagents > Two-component Reagents
HYDRANAL®
Two-component Reagents
Ordering Information
Composition
With two-component reagents, the reactants are in two separate solutions. HYDRANAL®-Solvent is a solution of sulfur dioxide and imidazole in methanol. HYDRANAL®-Titrant is a solution of iodine, dissolved in a suitable solvent, with an exact titre and a high stability of the titre.
Stability
The titrants have a shelf life of three years, the solvents of five years.
Advantages of the two-component titration
  • Higher titration speed compared with volumetric one-component titration.
  • Greater accuracy for small amounts of water.
  • Higher buffer capacity.
  • Exact and stable titre.
HYRDRANAL videos
Learn more - watch our
HYDRANAL videos
View Product List and Ordering Information
Back to top
Back to HYDRANAL®-home