Sigma-Aldrich
Analytical / Chromatography > Titration > ISE Equipment
Titration
ISE Equipment
Electrodes for ion-selective measurements (reference, ISE body, tool for removing membranes, PVC membrane)
Electrodes for ion-selective measurements.
NEW 16811 Reference electrode for ISE (Double-junction Ag/AgCl reference electrode, cable not included)
This reference electrode has specially developed for measuring with ion selective electrodes and can be used at very low ion concentrations. The electrode has two filling openings and a removable teflon diaphragm for easily exchanging the filling electrolyte.
Technical Specifications  
Reference electrolyte refillable, KCl c= 3 mol/L
Reference system Ag/AgCl cartridge, Ag+-free
Operating range 0…80 °C (32…176 °F)
Diaphragm teflon
Shaft material glass
Electrode plug S7
Application reference for ion-selective electrodes
Reference electrode for ISE.
45137 Electrode body for ISE
with Lemo connector for mounting of ion-selective membranes, cable (45153, 45136) and tool for removing membranes (44765) are not included.
Total length 140 mm
Diameter 12 mm
Shaft Polyoxymethylene (POM)
Electrode plug Lemo
 Electrode body for ISE.
NEWFluka 16613 Electrode Connecting Wire with S7/BNC-Connector for pH Electrodes (for Fluka 30948, 53162, 57458)
Electrode connecting wire, 1m coax. cable Electrode plug S7, BNC instrument plug used with pH meters of Beckmann, Corning, EDT, Fisher, Jenway, Hanna, Mettler-Toledo, Orion, Philips
NEWFluka 30676 Electrode Connecting Wire with S7/DIN-Connector for pH Electrodes (for Fluka 30948, 53162, 57458)
Electrode connecting wire, 1m coax. cable Electrode plug S7, DIN 19 262 instrument plug used with pH meters of Knick, Metrohm (Metrohm-pH meter <691), Schott, WTW
NEWFluka 49657 Electrode Connecting Wire with S7/Metrohm (LEMO)-Connector for pH Electrodes (for Fluka 30948, 53162, 57458)
Electrode connecting wire, 1m coax. cable Electrode plug S7, Metrohm (LEMO) instrument plug used with pH meters of Metrohm (pH meter ≥ 691)
NEWFluka 41734 Electrode Connecting Wire with S7/Banana-Connector for pH Electrodes (for Fluka 30948, 53162, 57458)
Electrode connecting wire, 1m coax. cable Electrode plug S7, Banana instrument plug
NEWFluka 11509 Adapter F-BNC Adapter for connection of BNC plugs (Fluka 16613, 45135)
to type F sockets used with pH meters of Metrohm (pH meter ≥ 691)
45135
Electrode Connecting Wire with Lemo/BNC-Connector for electrode body (Fluka 45137)
45136 Electrode Connecting Wire with Lemo/DIN-Connector for electrode body (Fluka 45137)
Back to top
Titration Home