Certipur® Reference Material for Reliable Calibration in pH Measurement
Reliable Calibration in pH Measurement
Accuracy of pH measurement results are directly affected by the buffer reference materials used in calibration. We offer a distinguished range of buffer solutions and substances for the exact calibration of your pH measurement equipment. Using Certipur® reference materials, you benefit from maximum accuracy, reliability and convenience and avoid costly repeat analyses.
For your strictest demands in equipment calibration and monitoring, we provide certified secondary standard buffer solutions and substances offering highest precision. For daily-routine calibration, we supply ready-to-use buffer solutions and Titrisol® buffer concentrates. Our ready-to-use buffer solutions in sealed sachets are ideal and convenient. Safely packed in small, handy quantities, they are always fresh and protected from contamination.
All our buffer solutions and substances for pH measurement calibration and monitoring are traceable to primary standards from PTB (Physical Technical Institute, Braunschweig, Germany) and from NIST (National Institute of Standards & Technolgy, Gaithersburg, USA). They are subjected to exhaustive quality controlled and qualified according ISO Guide 34 and measured in our DIN/EN ISO/IEC 17025 accredited calibration lab.