Sigma-Aldrich
Analytical / Chromatography > Vials & Accessories > Vials Taking 11mm Seals
Vials
Vials Taking 11mm Seals
Supelco offers crimp finish vials with two openings. We offer the standard vials with a 4.6 mm I.D. and also the large opening vial with a 6.0 mm opening. These vials are available in both clear and amber glass. The large opening vials are also offered with a marking spot on the side of the vial for sample identification.
11mm_finish
Vials only
Convenience packs
Micro-vials
Chromacol vials
Chromacol sleeves
Crimp seals with septa
Crimp seals (no septa)
Storage bin
Vials only (no seals)
Product Name Product # 
12 mm x 32 mm x 4.6 mm (I.D.)  
clear glass pack of 100 27068-U
clear glass pack of 1000 27069
amber glass pack of 100 27070-U
amber glass pack of 1000 27071-U
12 mm x 32 mm x 6.0 mm (I.D.)  
clear glass pack of 100 27058
clear glass pack of 1000 27059
amber glass pack of 100 27064
amber glass pack of 1000 27065
clear glass (with marking spot) pack of 100 27062-U
clear glass (with marking spot) pack of 1000 27063
amber glass (with marking spot) pack of 100 27066-U
amber glass (with marking spot) pack of 1000 27067-U
Convenience packs back to top
Multi-pak combinations of 2 mL large opening crimp top vials and 11 mm PTFE/red rubber aluminum seals.  
Product Name Product # 
clear glass pack of 100 27239
clear glass pack of 1000 27240-U
amber glass pack of 100 27241
amber glass pack of 1000 27242-U
Micro-vials (crimp top) back to top
Product Name Product # 
12 mm x 32 mm x 4.6 mm (I.D.)  
0.1 mL clear glass, conical pk of 12 33208
0.1 mL clear glass, conical pack of 144 33232
0.1 mL polypropylene, conical pack of 100 27075-U
0.1 mL polypropylene, conical pack of 1000 33217-U
0.1 mL plastic (with glass inner cone), conical pack of 100 27074-U
0.6 mL plastic (with glass inner cone), conical pack of 100 33255-U
12 mm x 32 mm x 6.0 mm (I.D.)  
1.5 mL clear glass Maximum Recovery pack of 100 27274
0.1 mL clear glass pack of 100 24714
W/0.3mL Insert amber glass pack of 100 27561-U
Chromacol vials (no seals) back to top
Product Name Product # 
Crimp Top Vial 0.9 mL clear glass vial , conical bottom pack of 125 27333
Crimp Top Vial 1.1 mL clear glass , tapered bottom O.D. × height 12 mm × 32 mm pack of 100 27310
Crimp Top Vial 1.1 mL clear glass O.D. × height 12 mm × 32 mm , tapered bottom pack of 1000 27311
Chromacol 1.7mL Clear Crimp Vial, 12 X 32 mm 1.7 mL clear glass body , crimp top pack of 1000 27309
Chromacol sleeves for 1.1 mL vials back to top
Product Name Product # 
Vial Support Sleeve, PTFE Support sleeve (non-disposable) O.D. × height 12 mm × 32 mm for use with 27310, 27311 pack of 50 27335
Sleeve for HP7673A 12 X 32 mm Chromacol Vial PTFE Support sleeve (non-disposable) for use with 27310, 27311 for HP7673A/1050A pack of 50 27336
Crimp seals (11 mm) with septa included back to top
Product Name Product # 
PTFE/rubber  
silver aluminum, KOMBO pack features seals in 12 colors, 25 of each color, pack of 300 27106
PTFE/red rubber (color coded seals)  
silver aluminum pack of 100 27102-U
silver aluminum pack of 1000 33233-U
red aluminum pack of 100 27103
red aluminum pack of 1000 27473
blue aluminum pack of 100 27104
blue aluminum pack of 1000 27381
gold aluminum pack of 100 27105
gold aluminum pack of 1000 27474
red PTFE/silicone  
silver aluminum pack of 100 27360-U
silver aluminum pack of 1000 27373
red PTFE/silicone/red PTFE  
silver aluminum pack of 100 27108
silver aluminum pack of 1000 27109-U
white PTFE/silicone  
silver aluminum pack of 100 27214-U
silver aluminum pack of 1000 27215
black Viton®  
silver aluminum pack of 100 27107
silver aluminum liner thickness .030 in. pack of 1000 33278-U
SPME applications  
aluminum, 5.6mm center hole, red PTFE/white silicone thickness 1.3 mm durometer: 35° shore A pack of 100 SU860094
aluminum, 5.6mm center hole, red PTFE/white silicone thickness 1.3 mm durometer: 35° shore A pack of 1000 SU860016
gold seal, magnetic, 5mm center hole, red PTFE/silicone/red PTFE thickness 1.0 mm durometer: 45° shore A pack of 100 SU860095
gold seal, magnetic, 5mm center hole, red PTFE/silicone/red PTFE thickness 1.0 mm durometer: 45° shore A pack of 1000 SU860017
gold seal, magnetic, 5mm center hole, red PTFE/white silicone thickness 1.3 mm durometer: 35° shore A pack of 100 SU860096
gold seal, magnetic, 5mm center hole, red PTFE/white silicone thickness 1.3 mm durometer: 35° shore A pack of 1000 SU860018
Crimp seals, 11 mm (no septa) back to top
Product Name Product # 
open center pack of 100 27099-U
Storage bin back to top
Product Name Product # 
Nine-Drawer Work Station filled with 3000 2 mL large opening crimp top clear glass vials, PTFE/red rubber 11 mm septa, and seals 27231-U