Vials
4mL Vials
Vials are 15 mm in diameter, 45 mm in height, and have a 13-425 thread.
• Choice of clear or amber glass.
• Precleaned or silanized options available
4ml
(product 27110 shown)
Vials only
Vials with caps
Vials with inserts
Kits
Caps
Septa
Inserts and springs
Vials only (screw top) back to top
Product Name Product # 
4 mL clear glass pack of 100 27111
4 mL clear glass pack of 1000 27031
4 mL amber glass pack of 100 27115-U
4 mL amber glass pack of 1000 27032
4 mL Silane-Treated clear glass pack of 100 27114
4 mL Silane-Treated clear glass pack of 1000 27220-U
4 mL Silane-Treated amber glasspack of 100 27217
4 mL Graduated clear glass pack of 100 27024
4 mL clear glass marking spot pack of 1000 27113
4 mL amber glass marking spot pack of 100 27116-U
4 mL amber glass marking spot pack of 1000 27117-U
2.5 mL Polypropylene Limited Volume Vial polypropylene volume 2.5 mL O.D. × height 15 mm × 45 mm pack of 100 27435
Vials with caps, precleaned and assembled back to top
Product Name Product # 
4 mL vial, 13-425 black phenolic hole cap  
clear glass pack of 100 27340
amber glass pack of 100 27345
Shell vial  
3 mL polypropylene, starburst conical snap plug (included) 27538
Vials with inserts back to top
Product Name Product # 
Snap-Top Vial, 4 mL clear glass vial and insert volume 350 µL pack of 100 27437
Snap-Top Vial, 4 mL, step amber glass, clear glass insert pack of 100 27438
Screw Top Vial (cap not included) 4 mL plastic body (with glass insert) pack of 100 27243
Kits back to top
Preassembled ( pack of 100)
Includes each of the following:
• screw top vials, 15 x 45 mm, 4 mL
• glass inserts, 0.30 mL clear glass, conical
• bottom springs for inserts
• closures, open top, phenolic, black
• septa, red PTFE/red rubber
Product Name Product # 
clear glass 27110
amber glass 27528
Caps, 13-425 back to top
Product Name Product # 
Hole caps  
black phenolic pack of 100 27120-U
black polypropylene pack of 100 27053
black polypropylene, PTFE/silicone thickness 0.060 in., preassembled pack of 100 27018
black top hat PTFE/silicone, preassembled pack of 100 27439
Solid caps  
green phenolic PTFE liner pack of 100 27141
black phenolic aluminum liner pack of 100 27142
Septa, 11 mm back to top
Product Name Product # 
Red PTFE/silicone/red PTFE thickness 0.040 in. pack of 100 27122-U
PTFE/rubber thickness 0.050 in. pack of 100 27145
Tan PTFE/Silicone thickness 0.060 in. pack of 100 27144
PTFE/silicone thickness 0.075 in. pack of 100 27356
PTFE/white silicone thickness 0.075 in. pack of 1000 27369-U
PTFE/Silicone thickness 0.060 in. pack of 100 27511
Barrier (aluminum/silicone) thickness 0.060 in. pack of 100 24883
Thermoseal ™ thickness 0.060 in. pack of 100 27192
Viton® thickness 0.060 in. pack of 100 27351
Cap Liner, 11 mm  
white PTFE thickness 0.010 in. pack of 100 27146
Inserts & springs for 15 x 45 mm vials back to top
Product Name Product # 
clear glass, shell style  
volume 0.75 mL diam. x height 8 mm x 43 mm pack of 200 24746
volume 0.75 mL diam. x height 8 mm x 43 mm pack of 1000 24747
clear glass, conical  
volume 0.25 mL diam. x height 6 mm x 39 mm pack of 100 27118-U
volume 0.25 mL diam. x height 6 mm x 39 mm pack of 1000 27076
volume 0.3 mL (with top spring) O.D. × height 6 mm × 38 mm pack of 100 27444
volume 0.275 mL (with plastic flange) O.D. × height 7 mm × 44 mm pack of 100 27443
volume 0.15 mL (with bottom spring) O.D. × height 6 mm × 38 mm pack of 100 24748
polypropylene, conical  
volume 0.15 mL (with bottom spring) O.D. × height 6 mm × 38 mm pack of 100 24749
volume 0.3 mL (with top spring) O.D. × height 6 mm × 38 mm pack of 100 27445
compression springs  
pack of 100 27086-U