Sigma-Aldrich
Analytical / Chromatography > Vials & Accessories > ABC Vials™
Vials
ABC Vials™
abc These universal 2 mL vials can be used with either crimp-top or screw-top closures. The shorter cap (9 mm thread) and longer neck allows them to be used with robotic systems. This wide opening (6.0 mm) 2 mL vial is 12 mm in diameter and 32 mm in height.
• 
40% wider opening than standard screw top vials
• 
Superior thread design ensures a secure seal
• 
Compatible with most GC autosamplers that accept crimp top vials
abc-vial-thread
shorter cap
longer neck
Preassembled
Vials only
Screw caps with septa
Inserts & springs
Preassembled ABC Vial™ back to top
Product Name Product # 
PTFE/silicone septum, clear polypropylene cap  
clear glass pack of 100 27487-U
clear glass pack of 1000 27542
clear glass (with marking spot) pack of 100 27490-U
clear glass (with marking spot) pack of 1000 27548-U
amber glass pack of 100 27493
amber glass (with marking spot) pack of 100 27496
amber glass (with marking spot) pack of 1000 27549-U
PTFE/red rubber septum, clear polypropylene cap  
clear glass pack of 100 27488-U
clear glass pack of 100 27544-U
clear glass (with marking spot) pack of 100 27491-U
clear glass (with marking spot) pack of 1000 27550-U
amber glass pack of 100 27494
amber glass vial (with marking spot) pack of 100 27497-U
amber glass (with marking spot) pack of 1000 27551
PTFE/silicone/PTFE septum, clear polypropylene cap  
clear glass pack of 100 27489
clear glass pack of 1000 27546-U
clear glass (with marking spot) pack of 100 27492-U
clear glass (with marking spot) pack of 1000 27552
amber glass pack of 100 27495
amber glass (with marking spot) pack of 100 27498
0.3 mL LVI clear glass vials with clear polypropylene cap  
PTFE/red rubber septum pack of 100 27564-U
PTFE/silicone septum pack of 100 27565-U
PTFE/silicone/PTFE septum pack of 100 27566-U
(no septum) pack of 100 27567-U
Vials only back to top
Product Name Product # 
clear glass pack of 100 27329
clear glass pack of 1000 27554
clear glass (with marking spot) pack of 100 27330
clear glass (with marking spot) pack of 1000 27556
amber glass pack of 100 27331
amber glass pack of 1000 27555-U
amber glass (with marking spot) pack of 100 27332
amber glass (with marking spot) pack of 1000 27557
0.3 mL LVI clear glass pack of 100 27562-U
0.3 mL LVI amber glass pack of 100 27563-U
Screw caps with septa back to top
Product Name Product # 
PTFE/silicone clear cap pack of 100 27326
PTFE/silicone clear cap pack of 1000 27558
PTFE/silicone/PTFE clear cap pack of 100 27328
PTFE/silicone/PTFE clear cap pack of 1000 27560-U
PTFE/silicone/PTFE black cap pack of 100 507407
PTFE/silicone/PTFE yellow cap pack of 100 507474
PTFE/silicone/PTFE red cap pack of 100 507431
PTFE/red rubber clear cap pack of 100 27327
PTFE/red rubber clear cap pack of 1000 27559
red PTFE/silicone black cap pack of 100 507393
red PTFE/silicone red cap pack of 100 507423
tan PTFE/silicone black cap pack of 100 507385
tan PTFE/silicone yellow cap pack of 100 507458
tan PTFE/silicone red cap pack of 100 507415
Inserts & springs back to top
Product Name Product # 
0.35 mL glass (shell style) height 31 mm pack of 100 24715
0.35 mL glass (shell style) height 31 mm pack of 1000 24716
0.25 mL glass conical height 31 mm pack of 100 24717
0.25 mL glass conical height 31 mm pack of 1000 24718
0.15 mL glass conical (with top spring) height 29 mm pack of 100 24719
0.15 mL glass conical (with top spring) height 29 mm pack of 500 24720
0.20 mL glass conical (with bottom spring) height 29 mm pack of 100 24721
0.20 mL polypropylene conical (with bottom spring) height 29 mm pack of 100 24722