Sigma-Aldrich
Analytical / Chromatography > Vials & Accessories > Accessories
Vials
Accessories
Many products are offered to accompany the vial products that we sell. We offer a range of vial racks, made of polypropylene material, to use to store your samples. In addition, safety-coated storage bottles, glass magnet products, Mini-Vaps, and Block Heaters are listed for usage in your application.
Vial racks click to see a picture of this item back to top
Economical and convenient storage for samples in your screw cap and crimp seal vials. Can be stacked when empty or full. Autoclavable, 120 °C.  
Product Name Product # 
for use with 6-8mm vial (96 vials per rack) pack of 5 23204-U
for use with 11-12mm vial (50 vials per rack) pack of 5 23207
for use with 15-16mm vial (48 vials per rack) pack of 5 23205-U
for use with 17mm vial (90 vials per rack) pack of 5 23202
for use with 21-23mm vial (36 vials per rack) pack of 5 23201
for use with 29mm vial (50 vials per rack, for 40mL EPA vial) pack of 5 23206
Storage bottles, plastic coated click to see a picture of this item back to top
•  Ideal for storing fluids, dry chemicals, and other lab materials
•  Slip resistant plastisol coating reduces potential breakage and protects lab personnel
•  Wide opening , special lip, and sloping shoulders make pouring and cleaning easier
•  Low alkali borosilicate gl ass content does not harm fluids
•  Autoclavable
Each bottle comes with a white polypropylene screw cap with a wedge-shaped inner sealing ring and polypropylene pour ring.
Product Name Product # 
100 mL clear glass O.D. × height 59 mm × 10 mm pack of 12 23171
500 mL clear glass O.D. × height 91 mm × 187 mm pack of 12 23172
1000 mL clear glass O.D. × height 104 mm × 224 mm pack of 12 23173
Cap for Mobile Phase Reservoir System GL 45 solid (no holes) pack of 12 23170-U
Storage bin click to see a picture of this item back to top
A convenient way to organize your vials. Available filled with supplies or unfilled.  
Product Name Product # 
Nine-Drawer Work Station filled with 2000 2 mL standard opening screw top clear glass vials, red PTFE/silicone septa, and black phenolic caps (recommended for Varian and Spectra Physics autosamplers) 27264-U
Nine-Drawer Work Station filled with 3000 2 mL large opening crimp top clear glass vials, PTFE/red rubber 11 mm septa, and seals 27231-U
Nine-Drawer Work Station unfilled (cabinet only) 27263
Glass magnets click to see a picture of this item back to top
Our Glass Magnet Sheet prevents your glass apparatus from being knocked over accidentally. This 2' x 2' sheet (61 cm x 61 cm), with its tacky surface, effectively secures the vessels in place. It can be cut with a razor blade or utility knife to fit strategically on counter tops or in drawers containing fragile glassware. Add it to carts when transporting glass to another lab or into the field. The Glass Magnet Vial Holder keeps individual vials, ampuls, and other vessels in place.  
Product Name Product # 
Glass Magnet mat width × length × thickness 24 in. × 24 in. × 3/16 in. pack of 1 57269
Glass Magnet diam. × thickness 4 in. × 3/16 in. pack of 2 57270
GLASS MARKING PEN 64800-U
2.5 mL vials, crimp caps and septa back to top
Product Name Product # 
2.5 mL vial (requires 13 mm seal) O.D. × height 11.7 mm × 34.5 mm pack of 288 33121
Crimp Seal (without septum) diam. 13 mm removable center pack of 100 27222-U
Crimp Seals with PTFE/Rubber Septa diam. 13 mm removable center pack of 100 27223
Crimp Seals with PTFE/Rubber Septa diam. 13 mm removable center pack of 1000 27451
Stoppers septum diam. 13 mm butyl rubber pack of 100 27224
Stoppers septum diam. 13 mm red rubber pack of 100 27226
Crimp Seals with PTFE/Silicone Septa diam. 13 mm pack of 100 27452
Special use vials back to top
Product Name Product # 
Shimadzu 5mL Screw Top Vial, 12 X 96mm, 10/425 Thread size 5 mL clear N-51A borosilicate glass body thread 10-425 27319-U
Shimadzu 5mL Screw Top Vial, 12 X 96mm, 10/425 Thread size 5 mL clear N-51A borosilicate glass body thread 10-425 27320-U
Shimadzu 5mL Vial, 12 X 96mm, PP Hole Cap with PTFE Septa Shimadzu 5mL vial and closure pre-assembled closure type, screw cap polypropylene caps (10-425) (PTFE Septa) 27321
Fisons 2.5mL Vial, 12 X 40mm size 2.5 mL O.D. × height 12 mm × 40 mm vial type, crimp top clear KG-33 borosilicate glass body 27323
Shimadzu 5mL Vial, 12 X 96mm size 5 mL O.D. × height 12 mm × 40 mm vial type, crimp top 27324
Shimadzu 5mL Vial, 12 X 96mm size 5 mL O.D. × height 12 mm × 40 mm clear KG-33 borosilicate glass body vial type, crimp top 27325