Sigma-Aldrich
Analytical / Chromatography > Vials & Accessories > Amber Bottles
Vials
Amber Bottles
These large amber screw top septum bottles will protect your sample from sunlight. Only glass and Teflon contact the contents.
amber_bottles
30 mL (32 x 80 mm)
60 mL (38 x 95 mm)
120 mL (49 x 114 mm)
240 mL (61 x 143 mm)
30 mL (32 x 80 mm) back to top
Product Name Product # 
This size bottle uses a 20-400 cap and 18 mm septum  
Bottle, black phenolic hole cap, PTFE/silicone septum pack of 12 23228
Bottle only pack of 12 23232
black phenolic hole cap (20-400) pack of 100 27176
PTFE/Silicone septum diam. x thickness 18 mm x 0.060 in. pack of 144 23242-U
black phenolic solid cap (20-400), with polyseal liner pack of 144 23238
60 mL (38 x 95 mm) back to top
This size bottle uses a 20-400 cap and 18 mm septum  
Bottle, black phenolic hole cap, PTFE/silicone septum pack of 12 23229
Bottle only pack of 12 23233
black phenolic hole cap (20-400) pack of 100 27176
black phenolic solid cap (20-400), with polyseal liner pack of 144 23238
PTFE/Silicone septum diam. × thickness 18 mm × 0.060 in. pack of 144 23242-U
120 mL (49 x 114 mm) back to top
This size bottle uses a 22-400 cap and 20 mm septum  
Bottle, black phenolic hole cap, PTFE/silicone septum pack of 12 23230-U
Bottle only pack of 12 23234
black phenolic hole cap (22-400) pack of 144 23236
black phenolic solid cap (22-400), with polyseal linerpack of 144 23240-U
tan PTFE(0.002 in.)/silicone septum O.D. × thickness 20 mm × .125 in. pack of 100 27236
PTFE(0.010 in.)/Silicone septum diam. × thickness 20 mm × .100 in. pack of 100 27237-U
240 mL (61 x 143 mm) back to top
This size bottle uses a 24-400 cap and 22 mm septum  
Bottle, black phenolic hole cap, PTFE/silicone septum pack of 12 23231
Bottle only pack of 12 23235
black phenolic hole cap (24-400) pack of 144 23237
black phenolic solid cap (24-400), with polyseal liner pack of 144 23241
PTFE/Silicone septum diam. × thickness 22 mm × 0.100 in. pack of 144 23245-U