Vials
Block Heater
Constant temperatures for derivatization and other reactions. The block heater features:
• Two temperature ranges, ambient to 60 °C, 50 °C to 130 °C
• Three-position power switch and two adjustable thermostats
• Pilot light shows heat cycling operation
Two models: 120 volts (100 watts, 0.83 amps, 50/60 Hz) and 240 volts (100 watts, 0.42 amps, 50/60 Hz).
Order blocks separately. Block 1 has eight holes (21 mm wide and 30 mm deep), and accepts our 3 mL or 5 mL micro reaction vessels. Block 2 has twelve holes (17 mm wide and 24 mm deep), and accepts our 0.1 mL to 2 mL micro reaction vessels.
block-heater
Block heater
(8-hole block not included)
8-hole-block
8-hole block for heater
Product Name Product # 
Block Heater AC input 110 V block (not included) 33315
Block Heater AC input 240 V CE compliant block (not included) 33318-U
Block #1 33316
Block #2 33317-U
Related products:
Micro Reaction Vessels