Vials
Headspace Vials
Headspace vials are available in volumes from 6 mL to 27 mL in clear glass. We offer vials with a flat top or a beveled top, and are available with either a flat bottom or a rounded bottom for your application.
headspace
Vials only
Chromacol
Convenience packs
Seals and septa for vials taking 20 mm seals
  Crimp seals with septa included
  Crimp seals only
  Septa
  Stoppers
Vials only back to top
Product Name Product # 
Beveled top, flat bottom, clear glass  
6 mL O.D. × height 23 mm × 38 mm pack of 100 27197
6 mL O.D. × height 22 mm × 38 mm pack of 100 27292
6 mL O.D. × height 22 mm × 38 mm pack of 1000 27293
10 mL O.D. × height 23 mm × 46 mm pack of 100 27198
20 mL O.D. × height 23 mm × 75 mm pack of 100 27199
Beveled top, rounded bottom, clear glass  
10 mL O.D. × height 22.6 mm × 46 mm pack of 100 27294
10 mL O.D. × height 22.6 mm × 46 mm pack of 1000 27295
20 mL O.D. × height 22.6 mm × 75 mm pack of 100 27296
20 mL O.D. × height 22.6 mm × 75 mm pack of 1000 27297
27 mL O.D. x height 30 mm x 60 mm pack of 100 27298
27 mL O.D. × height 30 mm × 60 mm pack of 1000 27299
Flat top, rounded bottom, clear glass  
10 mL O.D. × height 22.5 mm × 46 mm for use with Carlo Erba, CTC, Fisons, Varian (CP) pack of 100 854180-U
20 mL O.D. × height 22.5 mm × 75.5 mm for use with CTC PAL (Varian, Gerstel, Atas, Shimadzu) pack of 100 854181-U
Chromacol back to top
Product Name Product # 
Clear glass, beveled top, rounded bottom  
6 mL O.D. × height 22 mm × 38 mm , 6-CV pack of 100 508403
10 mL O.D. × height 22 mm × 45 mm , 10-CV pack of 100 508438
12 mL O.D. × height 18 mm × 65 mm , 12-CV pack of 100 508446
20 mL O.D. × height 22 mm × 75 mm , 20-CV pack of 100 508454
Convenience packs
back to top
Each pack contains 100 crimp top clear glass vials and 100 pressure release aluminum seals.  
Product Name Product # 
10 mL with PTFE/silicone 27303-U
10 mL with PTFE/butyl 27304-U
20 mL with PTFE/butyl 27305-U
20 mL with PTFE/silicone 27306
20 mL with PTFE/butyl 27307
Seals and septa for vials taking 20 mm seals  
back to serum bottles
Crimp seals, 20 mm, septa included back to top
Product Name Product # 
silver aluminum  
PTFE/Rubber pressure release (open center) pack of 100 27454-U
PTFE/Silicone pressure release (open center) pack of 100 27455-U
PTFE/red rubber removable center pack of 100 27228
PTFE/butyl faced thickness 3.0 mm durometer: 50° shore A, 10 mm hole, open center pack of 100 SU860011
PTFE(.002 in.)/white Silicone, open center pack of 100 27229
white PTFE/white Silicone open center pack of 100 27362
white PTFE/white Silicone open center pack of 1000 27375
PTFE/butyl open center pack of 100 27301-U
black Viton open center pack of 36 33146-U
black Viton open center pack of 100 27245
black Viton open center pack of 1000 27246
Magnetic (tin plated)  
PTFE/Silicone open center pack of 100 27300
Gold, 5mm hole PTFE/Butyl thickness 3.0 mm durometer: 50° shore A pack of 100 854178-U
Gold, 5mm hole white PTFE/blue silicone thickness 3.0 mm durometer: 35° shore A pack of 100 854179-U
Gold, 8mm hole blue PTFE/white silicone thickness 1.5 mm durometer: 60° shore A pack of 100 SU860053
Gold, 8 mm hole white PTFE/blue silicone thickness 3.0 mm durometer: 35° shore A pack of 100 SU860105
Gold, 8mm Hole, Black Viton® open center pack of 100 SU860052
Crimp seals, silver aluminum, 20 mm (no septa) back to top
Product Name Product # 
removable center pack of 100 27227-U
open center pack of 100 27230-U
tear-away pack of 100 27016
pressure release pack of 100 27200
Septa, 20 mm back to top
Product Name Product # 
Black Viton® thickness 0.030 in. pack of 100 27247
Black Viton® thickness 0.030 in. pack of 1000 27248-U
PTFE/butyl faced thickness 0.135 in. pack of 100 27201
Blue PTFE/white silicone thickness 0.030 in. pack of 100 27539
PTFE/Rubber thickness .125 in. pack of 100 27233
tan PTFE(.002 in.)/Silicone O.D. × thickness .125 in. pack of 100 27236
PTFE(.010 in.)/silicone thickness .100 in. pack of 100 27237-U
Barrier (aluminum/silicone) thickness .100 in. pack of 100 27189
PTFE/Silicone thickness .060 in. pack of 100 27541
PTFE(0.005 in.)/Silicone thickness .125 in. pack of 100 27361
PTFE(0.005 in.)/Silicone thickness .125 in. pack of 1000 27374
Thermosealä   thickness 0.060 in. pack of 100 27540-U
Stoppers, 20 mm back to top
Product Name Product # 
gray butyl pack of 100 27232
red rubber pack of 100 27234
silicone pack of 100 27235-U