Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Vials & Accessories > 2mL Large Opening Vials
Vials
2mL Large Opening Vials
Our wide opening 2 mL vial is 12 mm in diameter, 32 mm in height, and has a 6.0 mm opening. The thread on this vial is 10-425.
lgopening
Vials only
Vials with cap and setpa, preassembled
Caps
Septa
Inserts
Vials only (screw top) back to top
Product Name Product # 
clear glass  
2 mL pack of 100 27265
2 mL pack of 2000 27266
Step vial insert volume 250 µL pack of 100 27418
amber glass  
2 mL pack of 100 27267-U
2 mL step vial pack of 100 27406
Step vial insert volume 250 µL pack of 100 27419
polypropylene  
0.5 mL conical bottom pack of 100 27410
0.75 mL conical bottom pack of 100 27411
1 mL pack of 100 27269
1 mL pack of 1000 27270-U
Vials with cap and septa, preassembled back to top
Product Name Product # 
2 mL clear glass, 10-425 polypropylene hole cap  
red PTFE/silicone pack of 100 27531
PTFE/silicone/PTFE pack of 100 27533
PTFE/silicone w/slit pack of 100 27534
2 mL amber glass, 10-425 polypropylene hole cap  
red PTFE/silicone pack of 100 27532
PTFE/silicone/PTFE pack of 100 27535
PTFE/silicone w/slit pack of 100 27536
Caps, 10-425 back to top
Product Name Product # 
Solid caps  
black polypropylene aluminum liner pack of 100 27415
black polypropylene PTFE liner pack of 100 27416
Hole caps  
black polypropylene pack of 100 27271
black polypropylene pack of 1000 27272-U
black polypropylene (with larger hole) pack of 100 27401
yellow polypropylene pack of 100 27413
white polypropylene pack of 100 27414
Assembled black polypropylene cap and PTFE/silicone pack of 100 27273
Top seal  
clear polypropylene thickness 0.010 in. pack of 100 27417
Septa, 10 mm back to top
Product Name Product # 
Red PTFE/silicone/red PTFE thickness 0.040 in. pack of 100 27275
Red PTFE/silicone/red PTFE thickness 0.040 in. pack of 1000 27276
Red PTFE/white silicone thickness 0.060 in. pack of 100 27277
Red PTFE/white silicone thickness 0.060 in. pack of 1000 27278
Red PTFE/white silicone (with slit) thickness 0.040 in. pack of 100 27279
Red PTFE/white silicone thickness 0.040 in. pack of 1000 27280-U
Cap Liner, 11 mm  
white PTFE thickness 0.010 in. pack of 100 27281
white PTFE thickness 0.010 in. pack of 1000 27282
Inserts back to top
Product Name Product # 
0.35 mL glass (shell style) height 31 mm pack of 100 24715
0.35 mL glass (shell style) height 31 mm pack of 1000 24716
Insert for Shell Vial volume 0.05 mL polypropylene conical with bottom spring insert O.D. × height 6 mm × 25 mm pack of 100 27408
0.25 mL glass conical height 31 mm pack of 100 24717
0.25 mL glass conical height 31 mm pack of 1000 24718
0.15 mL glass conical (with top spring) height 29 mm pack of 100 24719
0.15 mL glass conical (with top spring) height 29 mm pack of 500 24720
0.20 mL glass conical (with bottom spring) height 29 mm pack of 100 24721
0.20 mL polypropylene conical (with bottom spring) height 29 mm pack of 100 24722
0.25 mL glass (with glass flange) for use with Step Vials pack of 100 27407
0.25 mL polypropylene (with plastic flange) for use with Step Vials pack of 100 27409