| Screw top |
|
| Clear glass, graduated with hole cap (phenolic black) |
back to top |
| Dimensions: Height with cap, depth from septum to bottom of cone. |
|
| Product Name |
Product # |
| 0.1 mL O.D. × height × depth 16.5 mm × 32 mm × 16 mm, 15-425 pack of 12 |
33289 |
| 0.3 mL O.D. × height × depth 16.5 mm × 34 mm × 23 mm, 15-42 pack of 12 |
33291 |
| 1.0 mL O.D. × height × depth 16.5 mm × 40 mm × 33 mm, 15-42 pack of 12 |
33293 |
| 2.0 mL O.D. × height × depth 16.5 mm × 58 mm × 48 mm, 15-425 pack of 12 |
33295 |
| 3.0 mL O.D. × height × depth 20.5 mm × 42 mm × 42 mm, 20-400 pack of 12 |
33297 |
| 5.0 mL O.D. × height × depth 20.5 mm × 61 mm × 58 mm, 20-400 pack of 12 |
33299 |
| |
|
| 10.0 mL (not graduated) O.D. × height 24.0 mm × 72 mm, 24-400 pack of 12 |
27479 |
| |
|
| Clear glass, graduated with solid cap (phenolic black) |
back to top |
| Dimensions: Height with cap, depth from septum to bottom of cone. |
|
| Product Name |
Product # |
| 0.1 mL O.D. × height × depth 16.5 mm × 32 mm × 16 mm, 15-425 pack of 12 |
27034 |
| 0.3 mL O.D. × height × depth 16.5 mm × 34 mm × 23 mm, 15-425 pack of 12 |
27035 |
| 1.0 mL O.D. × height × depth 16.5 mm × 40 mm × 33 mm, 15-425 pack of 12 |
27036 |
| 2.0 mL O.D. × height × depth 16.5 mm × 58 mm × 48 mm, 15-425 pack of 12 |
27037 |
| 3.0 mL O.D. × height × depth 20.5 mm × 42 mm × 42 mm, 20-400 pack of 12 |
27038-U |
| 5.0 mL O.D. × height × depth 20.5 mm × 61 mm × 58 mm, 20-400 pack of 12 |
27039 |
| |
|
| Amber glass with hole cap (phenolic black) |
back to top |
| Dimensions: Height without cap. |
|
| Product Name |
Product # |
| 1.0 mL O.D. × height 13 mm × 41 mm, 13-425 pack of 12 |
27475-U |
| 2.0 mL O.D. × height 20 mm × 41 mm, 20-400 pack of 12 |
27476 |
| 3.0 mL O.D. × height 20 mm × 47 mm, 20-400 pack of 12 |
27477-U |
| 5.0 mL O.D. × height 20 mm × 62 mm, 20-400 pack of 12 |
27478-U |
| |
|
| Crimp top |
|
Clear glass, (no seal)  |
back to top |
| Product Name |
Product # |
| 2.0 mL pack of 12 |
27377 |
| 5.0 mL pack of 12 |
27378 |
| |
|
| Accessories |
back to top |
| Product Name |
Product # |
| Magnetic Stir Bar for use with For 3.0-5.0mL vessels Teflon pack of 6 |
23227 |
| Magnetic Stir Bar for use with For 0.1-2.0mL vessels Teflon pack of 6 |
23226 |