Sigma-Aldrich
Analytical / Chromatography > Vials & Accessories > Micro Reaction Vessels
Vials
Micro Reaction Vessels
These vials are made of heavy wall borosilicate glass. They have a cone shape interior for easy removal of small samples. They are supplied with Teflon-faced rubber septum and cap. These products can be autoclaved or centrifuged. We also offer Mininert valves, replacement septa, and the septa inserter..
micro_reaction
Screw top
Clear glass, graduated with hole cap
Clear glass, graduated with solid cap
Amber glass with hole cap
Crimp top
Clear glass, (no seal)
Accessories
Screw top  
Clear glass, graduated with hole cap (phenolic black) back to top
Dimensions: Height with cap, depth from septum to bottom of cone.  
Product Name Product # 
0.1 mL O.D. × height × depth 16.5 mm × 32 mm × 16 mm, 15-425 pack of 12 33289
0.3 mL O.D. × height × depth 16.5 mm × 34 mm × 23 mm, 15-42 pack of 12 33291
1.0 mL O.D. × height × depth 16.5 mm × 40 mm × 33 mm, 15-42 pack of 12 33293
2.0 mL O.D. × height × depth 16.5 mm × 58 mm × 48 mm, 15-425 pack of 12 33295
3.0 mL O.D. × height × depth 20.5 mm × 42 mm × 42 mm, 20-400 pack of 12 33297
5.0 mL O.D. × height × depth 20.5 mm × 61 mm × 58 mm, 20-400 pack of 12 33299
10.0 mL (not graduated) O.D. × height 24.0 mm × 72 mm, 24-400 pack of 12 27479
Clear glass, graduated with solid cap (phenolic black) back to top
Dimensions: Height with cap, depth from septum to bottom of cone.  
Product Name Product # 
0.1 mL O.D. × height × depth 16.5 mm × 32 mm × 16 mm, 15-425 pack of 12 27034
0.3 mL O.D. × height × depth 16.5 mm × 34 mm × 23 mm, 15-425 pack of 12 27035
1.0 mL O.D. × height × depth 16.5 mm × 40 mm × 33 mm, 15-425 pack of 12 27036
2.0 mL O.D. × height × depth 16.5 mm × 58 mm × 48 mm, 15-425 pack of 12 27037
3.0 mL O.D. × height × depth 20.5 mm × 42 mm × 42 mm, 20-400 pack of 12 27038-U
5.0 mL O.D. × height × depth 20.5 mm × 61 mm × 58 mm, 20-400 pack of 12 27039
Amber glass with hole cap (phenolic black) back to top
Dimensions: Height without cap.  
Product Name Product # 
1.0 mL O.D. × height 13 mm × 41 mm, 13-425 pack of 12 27475-U
2.0 mL O.D. × height 20 mm × 41 mm, 20-400 pack of 12 27476
3.0 mL O.D. × height 20 mm × 47 mm, 20-400 pack of 12 27477-U
5.0 mL O.D. × height 20 mm × 62 mm, 20-400 pack of 12 27478-U
Crimp top  
Clear glass, (no seal) click to see a picture of this item back to top
Product Name Product # 
2.0 mL pack of 12 27377
5.0 mL pack of 12 27378
Accessories back to top
Product Name Product # 
Magnetic Stir Bar for use with For 3.0-5.0mL vessels Teflon pack of 6 23227
Magnetic Stir Bar for use with For 0.1-2.0mL vessels Teflon pack of 6 23226