Vials
Precleaned Vials
Prices have been reduced up to 50% on all Supelco precleaned sampling vials. We have been able to reduce cost and want to pass the savings along to our customers. We are offering the same high-quality glassware as we have always provided.
precleaned
Cleaned for water sampling according to US EPA 40 CFR 136, Guidelines for Establishing Test Procedures for the Analysis of Pollutants.
Vials are assembled with open-top screw caps and PTFE faced silicone septa. They are available in both clear and amber glass and sizes from 2 mL to 40 mL. Clear vials are made of Wheaton-33 low extractable borosilicate glass. Amber vials are Wheaton-320 borosilicate glass.
All vials are supplied in a shrink wrapped module to prevent contamination. Also included in the package are vial labels to record sampling data and documentation. A certificate of analysis click to see a sample of this document
(Level 3 documentation) is supplied with the 40 mL precleaned vial, and a certificate of compliance is provided for all other vials.
Precleaned Jars and Bottles
p000368
We also offer precleaned jars and bottles in clear and amber glass. These containers are cleaned according to US EPA protocol B. The sizes range from 4 oz. to 128 oz. and include a black polypropylene cap with 0.030’’ Teflon liner.
Vials with hole cap and septa included, assembled back to top
Product Name Product # 
clear glass  
2 mL phenolic black pack of 100 27339
4 mL phenolic black pack of 100 27340
7 mL phenolic black pack of 100 27341
15 mL phenolic black pack of 100 27342
22 mL phenolic black pack of 100 27343
40 mL polypropylene, green, 28x98mm pack of 72 23188
amber glass  
2 mL phenolic black pack of 100 27344
4 mL phenolic black pack of 100 27345
7 mL phenolic black pack of 100 27346
15 mL phenolic black pack of 100 27347
22 mL phenolic black pack of 100 27348
40 mL polypropylene, green, 28x98mm pack of 72 23189
Jars and bottles, cap included, precleaned back to top
clear glass  
4 oz, tall, wide mouth, cap 48-400 pack of 24 24551-U
8 oz, tall, wide mouth, cap 58-400 pack of 24 24552
16 oz, tall, wide mouth, cap 63-400 pack of 12 24553-U
32 oz, tall, wide mouth, cap 89-400 pack of 12 24554
32 oz, tall, narrow mouth, cap 33-400 pack of 12 24550-U
amber glass  
4 oz, short, wide mouth cap 38-400 pack of 12 24555
8 oz, short, wide mouth cap 45-400 pack of 12 24556
16 oz, short, wide mouth cap 53-400 pack of 12 24557-U
32 oz, short, wide mouth cap 53-400 pack of 12 24558
32 oz, short, narrow mouth cap 33-430 pack of 12 24559
80 oz jug, narrow mouth cap 38-430 pack of 6 24560
128 oz jug, narrow mouth cap 38-430 pack of4 24561