| These bottles are offered in sizes that range from 2.5 mL to 100 mL. The 5-100 mL vials require a 20 mm crimp seal to be used. They are available in both clear and amber glass.We also offer 20 mm crimp seals. |
 |
| |
|
| Amber glass |
back to top |
| Product Name |
Product # |
| 5 mL O.D. × height 25 mm × 40 mm pack of 36 |
33141-U |
| 5 mL O.D. × height 25 mm × 40 mm pack of 288 |
33142-U |
| 10 mL O.D. × height 25 mm × 50 mm pack of 36 |
33143 |
| 10 mL O.D. × height 25 mm × 50 mm pack of 288 |
33144 |
| Clear glass |
back to top |
| Product Name |
Product # |
| 2.5 mL (requires 13 mm seal) O.D. × height 11.7 mm × 34.5 mm pack of 288 |
33121 |
| 5 mL O.D. × height 25 mm × 40 mm pack of 36 |
33102-U |
| 5 mL O.D. × height 25 mm × 40 mm pack of 288 |
33103 |
| 10 mL O.D. × height 25 mm × 50 mm pack of 36 |
33104 |
| 10 mL O.D. × height 25 mm × 50 mm pack of 288 |
33105-U |
| 30 mL O.D. × height 36.6 mm × 62.8 mm pack of 36 |
33106 |
| 30 mL O.D. × height 36.6 mm × 62.8 mm pack of 288 |
33107 |
| 50 mL O.D. × height 43 mm × 73 mm pack of 36 |
33108-U |
| 50 mL O.D. × height 43 mm × 73 mm pack of 272 |
33109-U |
| 100 mL O.D. × height 51.7 mm × 94.5 mm pack of 36 |
33110-U |
| 100 mL O.D. × height 51.7 mm × 94.5 mm pack of 144 |
33111-U |