Sigma-Aldrich
Analytical / Chromatography > Vials & Accessories > Serum Bottles
Vials
Serum Bottles
These bottles are offered in sizes that range from 2.5 mL to 100 mL. The 5-100 mL vials require a 20 mm crimp seal to be used. They are available in both clear and amber glass.We also offer 20 mm crimp seals.
serum_bottle
Amber glass back to top
Product Name Product # 
5 mL O.D. × height 25 mm × 40 mm pack of 36 33141-U
5 mL O.D. × height 25 mm × 40 mm pack of 288 33142-U
10 mL O.D. × height 25 mm × 50 mm pack of 36 33143
10 mL O.D. × height 25 mm × 50 mm pack of 288 33144
Clear glass back to top
Product Name Product # 
2.5 mL (requires 13 mm seal) O.D. × height 11.7 mm × 34.5 mm pack of 288 33121
5 mL O.D. × height 25 mm × 40 mm pack of 36 33102-U
5 mL O.D. × height 25 mm × 40 mm pack of 288 33103
10 mL O.D. × height 25 mm × 50 mm pack of 36 33104
10 mL O.D. × height 25 mm × 50 mm pack of 288 33105-U
30 mL O.D. × height 36.6 mm × 62.8 mm pack of 36 33106
30 mL O.D. × height 36.6 mm × 62.8 mm pack of 288 33107
50 mL O.D. × height 43 mm × 73 mm pack of 36 33108-U
50 mL O.D. × height 43 mm × 73 mm pack of 272 33109-U
100 mL O.D. × height 51.7 mm × 94.5 mm pack of 36 33110-U
100 mL O.D. × height 51.7 mm × 94.5 mm pack of 144 33111-U