Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Vials & Accessories > Silane Treated Vials
Vials
Silane Treated Vials
Proprietary silane treatments are available to shield active silanol groups on untreated glass surfaces. We have available as a stock product, many vial styles conditioned by this method. We also can treat other sizes as a custom order for your use in your laboratory.
silane-treated 
Product Name Product # 
Screw Top Vial, 2 mL clear glass pack of 100 27029-U
Screw Top Vial, 2 mL clear glass vial pack of 1000 27030
Screw Top Vial, 2 mL amber glass vial pack of 100 27238
Screw Top Vial, 2 mL amber glass vial pack of 1000 27219
Crimp Top Vial, 2 mL clear glass large opening, (6 mm) pack of 100 27060-U
Crimp Top Vial, 2 mL clear glass large opening, (6 mm) pack of 1000 27061
Crimp Top Vial, 2 mL amber glass large opening, (6 mm) pack of 100 27225-U
Crimp Top Vial, 2 mL amber glass large opening, (6 mm) pack of 1000 27216
Screw Top Vial, 4 mL clear glass pack of 100 27114
Screw Top Vial, 4 mL clear glass pack of 1000 27220-U
Screw Top Vial, 4 mL amber glass pack of 100 27217