Sigma-Aldrich
Analytical / Chromatography > Vials & Accessories > Snap-Top Vials
Vials
Snap-Top Vials
Vials are offered in two sizes, 2 mL and 4 mL. They are designed to be used with a plastic snap top cap. These caps eliminate the need for crimping. The caps simply snap over the top of the vial, sealing the contents safe inside.
snaptop
Vials only, large opening
2 mL vials with inserts
Snap-Top seals for 2 mL vials
Snap-Caps for 2 mL vials (tighter seal)
4 mL vials only
Snap-Top seals for 4 mL vials
Vials only, large opening (6.0 mm)
Product Name Product # 
clear glass  
2 mL pack of 100 24750-U
2 mL pack of 2000 24751
2 mL (with marking spot) pack of 100 27249
2 mL (with marking spot) pack of 1000 27250-U
amber glass  
2 mL pack of 100 24752
2 mL pack of 2000 24753
2 mL (with marking spot) pack of 100 27251
2 mL (with marking spot) pack of 1000 27252
polypropylene  
0.75 mL pack of 100 27253
0.75 mL pack of 1000 27254
0.50 mL pack of 100 27422
0.10 mL pack of 100 27423
2 mL large opening vials with inserts back to top
Product Name Product # 
amber plastic vial with 100 µL glass insert pack of 100 27425
Snap-Top seals for 2 mL vials back to top
Product Name Product # 
polypropylene  
PTFE pack of 100 24754
PTFE pack of 1000 24755
PTFE/red rubber pack of 100 24756
PTFE/red rubber pack of 1000 24757
PTFE/silicone pack of 100 24758
PTFE/silicone pack of 1000 24759
PTFE/silicone/PTFE pack of 100 24760-U
PTFE/silicone w/slit pack of 100 27426
PTFE/silicone (with starburst) pack of 100 27427
PTFE/silicone/PTFE (with starburst) pack of 100 27428
Viton® (with starburst) pack of 100 27429-U
polyethylene  
linerless pack of 100 24762
linerless pack of 1000 24763
Snap-Caps for 2 mL vials (tighter seal) back to top
Product Name Product # 
polypropylene, PTFE/rubber pack of 100 27430-U
polypropylene, PTFE/silicone pack of 100 27432
4 mL vials only (no caps) back to top
Product Name Product # 
clear glass pack of 100 27440-U
amber glass pack of 100 27441
Snap-Top seals (13 mm) for 4 mL vials back to top
Product Name Product # 
polypropylene  
PTFE-faced silicone starburst pack of 100 27449
PTFE/silicone/PTFE starburst pack of 100 27450-U