Sigma-Aldrich
Analytical / Chromatography > Vials & Accessories > SPME Specialty Vials
Vials
SPME Specialty Vials
A selection of specialty vials for SPME is available for your application in both screw cap and crimp top designs.
spme
We suggest you use a thinner septum for SPME applications to extend the life of your needle. We offer a series of vials with a thicker lip (Thin Seal) to maintain a tight seal, . The vials, seals and septa are available in 10 and 20 mL versions.
spme_vial_lip
thicker lip standard lip
Thin Seal - crimp top
Thin Seal - screw cap
Vials - screw top preassembled
Vials only - screw top
Screw caps
Accessories
Thin Seal - crimp top
Product Name Product # 
Thin Seal Vials (lip thicknes 0.125 in.)  
10 mL clear glass diam. × height 24.5  × 50 mm pack of 36 27385
10 mL clear glass diam. × height 24.5  × 50 mm pack of 144 27386
20 mL clear glass diam. x height 22.5 x 75.5 mm pack of 100 SU860104
Seals, 20 mm  
magnetic gold seal (with 8 mm center hole), black Viton® septum thickness 0.04 in. (1 mm) durometer: 75° shore A pack of 100 SU860106
magnetic gold seal (with 8 mm center hole), PTFE/silicone septum thickness 0.06 in. (1.5 mm) durometer: 60° shore A pack of 100 SU860053
aluminum seal open center, black Viton® septum thickness 0.030 in. pack of 36 33146-U
aluminum seal open center, black Viton® septum thickness 0.030 in. pack of 100 27245
aluminum seal open center, black Viton® septum thickness 0.030 in. pack of 288 28298-U
aluminum seal open center, black Viton® septum thickness 0.030 in. pack of 1000 27246
Septa, 20 mm  
blue PTFE/white silicone thin seal thickness 0.030 in. pack of 100 27539
Thin Seal - screw cap back to top
Product Name Product # 
10 mL vials, 22.5 mm × 46 mm, 18-400  
clear glass round bottom, for use with CTC PAL (Varian, Gerstel, Atas, Shimadzu) pack of 100 SU860099
amber glass round bottom, for use with CTC PAL (Varian, Gerstel, Atas, Shimadzu) pack of 100 SU860100
20 mL vials, 22.5 mm × 75.5 mm, 18-400  
clear glass round bottom, for use with CTC PAL (Varian, Gerstel, Atas, Shimadzu) pack of 100 SU860097
amber glass round bottom, for use with CTC PAL (Varian, Gerstel, Atas, Shimadzu) pack of 100 SU860098
Screw caps, 18-400  
magnetic with hole, PTFE-faced silicone septa, 35 Shore A thickness 1.3 mm pack of 100 SU860101
magnetic with hole, PTFE-faced butyl septa, 55 Shore A thickness 1.6 mm pack of 100 SU860102
magnetic with hole, PTFE/silicone septa, 60 Shore A thickness 1.5 mm pack of 100 SU860103
Vials - screw top, preassembled (for use with Varian 8200 autosampler) back to top
Product Name Product # 
with hole cap  
2 mL clear glass diam. × height 12 × 32 mm red PTFE/silicone black polypropylene cap (large opening), 10-425 pack of 100 27531
2 mL amber glass diam. × height 12 × 32 mm red PTFE/silicone black polypropylene cap (large opening), 10-425 pack of 100 27532
10 mL clear glass diam. × height 24.5  × 50 mm PTFE/silicone, phenolic, 22-400 pack of 100 27529
10 mL amber glass diam. × height 24.5  × 50 mm PTFE/silicone, phenolic, 22-400 pack of 100 27530
Vials only - screw top (no caps) back to top
Product Name Product # 
2 mL, 12 mm × 32 mm 10-425  
clear glass pack of 100 27265
amber glass pack of 100 27267-U
10 mL, 24.5 mm × 52 mm 22-400  
clear glass pack of 36 27387
clear glass pack of 144 27388
amber glass pack of 36 27389
amber glass pack of 144 27390-U
Screw caps back to top
Product Name Product # 
black polypropylene with hole for use with 2 mL SPME vial (10-425 thread) pack of 100 27273
black phenolic with hole for use with 10 mL SPME vial (22-400 thread) pack of 100 23236
Accessories back to top
Product Name Product # 
Acrylic Vial Rack length × width × height 160 mm × 160 mm × 30 mm, for 24 mm screw top vials, 25 cavities 1ea SU860107