Sigma-Aldrich
Analytical / Chromatography > Vials & Accessories > 2mL Standard Opening Vials
Vials
2mL Standard Opening Vials
Our standard 2 mL vial is 12 mm in diameter, 32 mm in height, and has a 4.6 mm opening. The thread size on this vial is 8-425.
• Choice of clear or amber glass.
• Precleaned or silanized options available
stdopening-screwcap
Vials only
Vials with cap and septa
Shell vials with plug
Micro-vials
Caps
Septa
Inserts
Storage bin
Vials only (screw top) back to top
Product Name Product # 
1.5 mL clear glass O.D. × height 12 mm × 30 mm pack of 100 507369
1.5 mL clear glass O.D. × height 12 mm × 30 mm pack of 1000 507377
2 mL clear glass pack of 100 27078
2 mL clear glass pack of 1000 27079
2 mL amber glass pack of 100 27083-U
2 mL amber glass pack of 1000 27087-U
Silane-treated  
2 mL clear glass pack of 100 27029-U
2 mL clear glass pack of 1000 27030
2 mL amber glass pack of 100 27238
2 mL amber glass pack of 1000 27219
Graduated  
2 mL clear glass pack of 100 27023
with marking spot  
2 mL clear glass pack of 100 27080-U
2 mL clear glass pack of 1000 27081-U
2 mL amber glass pack of 100 27084-U
2 mL amber glass pack of 1000 27085-U
Chromacol  
1.1 mL clear glass, 1.1-STVG pack of 100 27317
Polypropylene  
0.75 mL pack of 100 24709
0.75 mL pack of 1000 24710
Vials with cap and septa, preassembled and precleaned back to top
Product Name Product # 
2 mL vial, 8-425 black phenolic hole cap  
clear glass, PTFE/silicone pack of 100 27339
amber glass, PTFE/silicone pack of 100 27344
Shell vials with plugs back to top
Product Name Product # 
2 mL clear glass, TITESEAL (included) pack of 200 27255
2 mL clear glass, TITESEAL (included) pack of 2000 27256
2 mL amber glass, TITESEAL (included) pack of 2000 27258
Micro-vials back to top
Product Name Product # 
0.1 mL clear glass pack of 12 33206-U
0.1 mL clear glass pack of 144 33241
0.1 mL natural polypropylene pack of 200 24712
0.1 mL natural polypropylene pack of 1000 24713
0.1 mL plastic vial glass insert pack of 100 27090-U
0.1 mL amber plastic glass insert pack of 100 27412
Caps, 8-425 back to top
Product Name Product # 
Solid caps  
green phenolic PTFE liner pack of 100 27091-U
black phenolic aluminum liner pack of 100 27092-U
Hole caps  
black phenolic pack of 100 27094-U
black phenolic PTFE/silicone septum thickness 0.060 in. pack of 100 27093-U
black phenolic PTFE/silicone septum thickness 0.060 in. pack of 100 27093-U
black polypropylene pack of 100 27052
black polypropylene pack of 1000 24764
yellow polypropylene pack of 1000 24765
black polypropylene (with larger hole) pack of 100 27401
black polypropylene top hat (with larger hole) silicone septum pack of 100 27403-U
yellow polypropylene top hat (with larger hole) silicone septum pack of 100 27404
black polypropylene open-top Chromacol, 8-SC pack of 100 507644
white polypropylene pack of 100 507660
blue polypropylene cap of 100 507687
black polypropylene PTFE/Silicone Septum pack of 100 27262
clear polypropylene KIM-LOK PTFE/silicone septum pack of 100 27261-U
black polypropylene PTFE/silicone septum thickness 0.60 in. pack of 1000 33242
Septa, 8 mm back to top
Product Name Product # 
Red PTFE/white silicone/red PTFE thickness 0.040 in. pack of 100 27096-U
Red PTFE/white silicone/red PTFE thickness 0.040 in. pack of 1000 23182-U
Tan PTFE/white silicone thickness 0.060 in. pack of 100 27095-U
Tan PTFE/white silicone thickness 0.060 in. pack of 1000 33213
Red PTFE/white silicone thickness 0.060 in. pack of 100 27097-U
Red PTFE/white silicone thickness 0.060 in. pack of 1000 23243
Red PTFE/white silicone (with slit) thickness 0.060 in. pack of 100 27098-U
Red PTFE/white silicone (with slit) thickness 0.060 in. pack of 1000 24881
PTFE/red rubber thickness 0.050 in. pack of 100 27132
PTFE/Silicone thickness 0.060 in. pack of 100 27510-U
Black Viton® thickness 0.060 in. pack of 100 27350-U
Barrier (aluminum/silicone) thickness 0.060 in. pack of 100 24882-U
Thermosealä thickness 0.060 in. pack of 100 27191
Cap Liner, 9 mm  
white PTFE thickness 0.010 in. pack of 100 27133
Inserts back to top
Product Name Product # 
0.25 mL clear glass (shell style) O.D. × height 5 mm × 31 mm pack of 200 24701
0.25 mL clear glass (shell style) O.D. × height 5 mm × 31 mm pack of 1000 24702
0.10 mL clear glass conical O.D. × height 5 mm × 31 mm pack of 200 24703
0.10 mL clear glass conical O.D. × height 5 mm × 31 mm pack of 1000 24704
Self-Centering Support for 24704 Insert pack of 500 27338
0.15 mL clear glass conical (with polyflange) O.D. × height 5 mm × 29 mm pack of 100 24705
0.15 mL glass insert conical polyflange O.D. × height 5 mm × 29 mm pack of 500 24706
0.05 mL glass insert conical bottom spring O.D. × height 4 mm × 25 mm pack of 100 27400-U
0.15 mL clear glass conical (with bottom spring) O.D. × height 5 mm × 29 mm pack of 100 24707
0.15 mL polypropylene conical (with bottom spring) O.D. × height 5 mm × 30 mm pack of 100 24708
Storage bin back to top
Product Name Product # 
Nine-Drawer Work Station filled with 2000 2 mL standard opening screw top clear glass vials, red PTFE/silicone septa, and black phenolic caps (recommended for Varian and Spectra Physics autosamplers) 27264-U