Sigma-Aldrich
Analytical / Chromatography > Videos & Web Seminars > HYDRANAL Videos > Coulometric Titration, Cell with Diaphragm
HYDRANAL® Videos
Coulometric Titration, Cell with Diaphragm
Karl Fischer Video on Coulometric Titration using HYDRANAL®-Coulomat Reagents in a Coulometric Cell with a Diaphragm.
The various titration steps for this method are described below the video, also a representation of the typical coulometric cell with a diaphragm.
| View Products | Multimedia Guide on CD | Questions / Comments |
Analytical Seminars
Sign up to attend a live seminar!
Titration Steps:
Filling the anodic compartment
(with HYDRANAL®-Coulomat A/AG/AD/AG-H/AG-Oven/AK/Oil/E)
If the cell is dismantled, it must be dried at 50°C in a drying cabinet prior to use. The drying of the diaphragm requires special care. A slight excess of iodine present during the manufacture of HYDRANAL-Coulomat A and E types removes traces of moisture from the reagent. This solution is added to the anodic compartment of the coulometric titration cell using a dry funnel. The amount of reagent used (usually 100-150 mL) depends on the instrument used. The titration cell is closed immediately after the addition of reagent.
Filling the cathodic compartment
(with HYDRANAL®-Coulomat CG/CG-K or HYDRANAL®-Coulomat E)
The cathodic compartment is filled with HYDRANAL-Coulomat CG/CG-K/E (usually 5 mL). The level of the catholyte in the cathodic compartment should be lower than that of the anolyte in order to prevent diffusion into the anodic compartment. The cathodic compartment must also be anhydrous.
Drying the titration cell
The titration cell is self-drying. The instrumentation determines and indicates the degree of dryness automatically as the "drift" or "background" in µg/min or µg/sec of water. The drift value for a freshly replenished titration cell should not exceed 10 µg/min (ideally, 4 µg/min) of water. The drift can increase during a series of determinations. As long as the drift rate is stable, this does not appreciably influence the accuracy of the analyses since the instrumentation automatically compensates for the inherent drift value. Slow release of moisture that adheres to the walls can cause drift - this residual moisture can be quickly removed by gently swirling the filled titration vessel. Sometimes, moisture from the cathodic compartment can also result in high drift. Reductive substances produced in a catholyte, spent or unsuitable, have the same effect; hence, it is recommended that the catholyte be replaced at least once a week.
Addition of the sample
Titration is initiated by pressing the start button. The liquid sample to be analyzed is then injected through the septum into the anolyte. Solid samples, dissolved in suitable solvents, are added in dissolved form. Also, the moisture can be evaporated from the solid sample in an oven, collected and added as vapor to the titration cell. Gaseous samples are introduced into the anolyte via a gas inlet tube. The size of the sample chosen depends on the anticipated water content and the desired accuracy. The sample size should not exceed 10 mL for practical reasons. The coulometric titration cell will hold approximately 50 mL – a single titration of 10 mL of sample means a total of 5 determinations possible per reagent fill. Generally, 0.5-5.0 mL samples in liquid form are administered. Gaseous samples range from 100 mL to 10 L.
Calculation of the results
Coulometric titration equipment indicates the amount of water determined digitally in mg or µg. Most instruments will show the results in mass or volume units (% or ppm) if the sample size was entered.
Replacement of the reagents
coulomat_diaphragm The spent reagent can be removed from the anodic cell with a plastic suction bottle; certain instruments are equipped with pumps. The anodic compartment is subsequently refilled with HYDRANAL-Coulomat A/AG/AD/AG-H/AG-Oven/AK/Oil/E, and the cathodic compartment with HYDRANAL-Coulomat CG/CG-K/E. Determining when the reagent is spent or requires changing is not always obvious. Usually, a full titration cell necessitates replacement of the reagent. Also, successive dilution reduces the conductivity and necessitates replenishment of the reagent.
The capacity limit of HYDRANAL-Coulomat (anolyte) is seldom reached. The capacity of the catholyte must be determined empirically.
A drift increase often means the reagent must be replaced. An increase in the drift can have different causes. In case of doubt, the recovery rate of water standard should be checked - this method is a reliable way of determining whether the equipment is functioning properly.
Performance Tip
The coulometric cell should always be dry and operational. This can be achieved if the machine remains permanently switched on. Moisture penetrating the cell will be immediately eliminated this way. If the machine is switched off, water collects in the cell; after start up, water elimination takes up time and reagent.