Analytical / Chromatography > Videos > SPME Videos
Analytical / Chromatography Videos
SPME Videos
spme_center
SPME is a unique sample preparation technique that eliminates most drawbacks to extracting organics. SPME requires no solvents or complicated apparatus. It can concentrate volatile and nonvolatile compounds (in liquids or gaseous samples), for subsequent analysis by GC or HPLC.
SPME offers some important advantages:
  • Single Step Extraction - reduces sample preparation time by 70%
  • Versatility - various selectivities available, adapts to any GC or HPLC system, can be automated
  • Efficient & Economical - little/no use of solvents, quantitative method, allows more than 50 extractions per fiber on average
View our SPME video selection below to see how the SMPE technique can meet your sample preparation needs.
  Camera_spools Introduction to SPME   Camera_spools Auto SPME Sampling
  Camera_spools Solventless Sample Prep   Camera_spools Headspace Sampling with SPME
  Camera_spools Portability   Camera_spools Direct Immersion Sampling
  Camera_spools Installing an SPME Fiber   Camera_spools Getting Started with SPME
  Camera_spools Manual SPME Sampling   Camera_spools Reusability
Order SPME products through our online product catalog.
Brazil Video com tradução para Português