Analytical / Chromatography > Videos & Web Seminars > SPME Videos > Headspace Sampling of Volatiles
SPME Videos
Headspace Sampling of Volatiles
Brief video demonstrates the use of a SPME fiber for headspace sampling of volatile organic compounds (VOCs), followed by injection into a GC system for chromatography analysis.
Analytical Seminars
Sign up to attend a live seminar!
| View Products | Request SPME CD |
| Video Feedback |
video with Brazil Portuguese Vídeo com tradução para Português
Headspace sampling with SPME is the preferred method to extract volatile organic compounds. Begin the extraction process by placing the sample into the vial puck on the hotplate and index the vial to the sampling position. Insert the holder into the sampling stand, passing the needle through the septum that seals the sample vial. When sampling a liquid, position the SPME fiber in the headspace just above the stirring vortex of the liquid sample. If a solid sample is to be extracted, position the SPME fiber above the surface of the solid without touching it. Retract the fiber into the protective needle and transfer it to the GC injection port.