Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
VersaFlash™ Videos
Quick Cartridge Change-out
Video clip demonstrates the simplicity, speed and ease with which a VersaPak® cartridge can be removed and replaced on the VersaFlash® station of our flash chromatography system.
Analytical Seminars
Sign up to attend a live seminar!
| VersaFlash Products | Request VersaFlash CD |
| Video Feedback |
video with Brazil Portuguese Vídeo com tradução para Português
One of the unique features of the VersaFlash system is how quick and easy it is to change out cartridges between samples. Once you’ve processed the first sample, simply turn the handle on the VersaFlash station counterclockwise to the “idle” position and pull it out to release the spent cartridge. Then simply replace the spent cartridge with a new cartridge, push the handle in, and turn the handle clockwise to the “operate” position to seal the cartridge into the VersaFlash station. When working with the same size cartridges, cartridge change-out is fast and simple.