Sigma-Aldrich
Analytical / Chromatography > Videos & Web Seminars > VersaFlash Videos > Direct Sample Loading
VersaFlash™ Videos
Direct Sample Loading
Video demonstrates direct sample loading onto the flash chromatography cartridge using a manual syringe.
Analytical Seminars
Sign up to attend a live seminar!
| VersaFlash Products | Request VersaFlash CD |
| Video Feedback |
video with Brazil Portuguese Vídeo com tradução para Português
The fittings on the cartridge end caps are female luer fittings; therefore, it is very easy to attach a syringe pre-filled with the sample into the end cap. Then simply inject the sample onto the cartridge and remove the syringe. Be careful when direct-loading a sample with a syringe when the cartridge has been preconditioned with solvent, as the sample will displace solvent in the cartridge.