Analytical / Chromatography > Videos & Web Seminars > VersaFlash Videos > Rev-Elution (Back-Flushing)
VersaFlash™ Videos
Rev-Elution (Back-Flushing)
Video demonstrates the elution of sample impurities using an appropriate solvent in the first step, followed by reversal of the cartridge and back-flushing the clean sample (reversing direction of elution) in the next step.
Analytical Seminars
Sign up to attend a live seminar!
| VersaFlash Products | Request VersaFlash CD |
| Video Feedback |
video with Brazil Portuguese Vídeo com tradução para Português
Researchers commonly encounter samples where the best way to purify the sample is to choose a solvent system that leaves the target compound on the origin, running off any interfering impurities. Once the impurities are gone, the symmetric VersaPak cartridge design allows you to flip the cartridge over so that the target compound is now near the cartridge outlet. By changing the solvent system, you can more quickly elute the target compound without having to wait for the compound to move through the entire cartridge bed.