Sigma-Aldrich
Analytical / Chromatography > Videos & Web Seminars > VersaFlash Videos > Solid Sample Cartridge Loading
VersaFlash™ Videos
Solid Sample Cartridge Loading
Video shows how solid samples or low-solubility samples can be loaded into an empty cartridge to enable a flash chromatography separation on the VersaFlash® system.
Analytical Seminars
Sign up to attend a live seminar!
| VersaFlash Products | Request VersaFlash CD |
| Video Feedback |
video with Brazil Portuguese Vídeo com tradução para Português
Still another way to load samples is to use a solid sample cartridge. Some samples that have low solubility in the solvent or solid samples such as natural products can be dried loaded into an empty cartridge. The common process for the solid sample loading is to deposit a sample on an inert support like silica or celite by rotovapping a slurry of sample and support. Then fill the solid sample cartridge with the sample, install the disposable frit using the frit insertion tool. Remove the frit insertion tool and attach the stainless steel cap. Install the solid sample cartridge in series with the separation cartridge as shown in the cartridge-stacking segment. The sample is then leached onto the separation cartridge. The solid sample cartridge can also be used to custom-pack novel resins and media to create unique separation modes. This enables the researcher to create chromatographic separations matched to their specific applications. These cartridges can be used individually as purification columns or can be stacked with other purification columns to create bi-modal separations.
ORDERING
SERVICE & SUPPORT
CORPORATE
    Follow us on Twitter!  Join us on Facebook!  Join us on LinkedIn!  Watch our videos on YouTube!
Copyrights © 2010 Sigma-Aldrich Co. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.