VersaFlash™ Videos
Symmetric End-Fittings
Video shows how the unique design of the symmetric VersaPak® flash cartridge adds flexibility to the VersaFlash® flash chromatography system.
Analytical Seminars
Sign up to attend a live seminar!
| VersaFlash Products | Request VersaFlash CD |
| Video Feedback |
video with Brazil Portuguese Vídeo com tradução para Português
What makes the VersaFlash system so flexible and versatile is the unique design of the VersaPak cartridges. The VersaPak cartridges have a thick wall which eliminates the need for compression barrels or other supporting hardware. The cartridges are completely symmetric with the end fittings at both ends identical. This means that technically there’s no top or bottom or inlet or outlet to the cartridges. Samples can be loaded at either end of the cartridge and processed in many different ways. The symmetric end fittings also lead to the ability to stack cartridges, reverse cartridges, quickly change out and scale up cartridges.