Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
VersaFlash™ Videos
Upward / Downward Flow
Video illustrates the easy set up of the flash chromatography system for upward flow, as may be desired for some separations.
Analytical Seminars
Sign up to attend a live seminar!
| VersaFlash Products | Request VersaFlash CD |
| Video Feedback |
video with Brazil Portuguese Vídeo com tradução para Português
With pre-packed VersaPak cartridges and the VersaFlash station, you can use downward flow or upward flow. Some researchers feel that upward flow leads to more thorough expulsion of air from the cartridge, minimizing the potential for channeling or voiding, which could adversely affect the quality of the separation.