Sigma-Aldrich
Analytical / Chromatography > Videos & Web Seminars > VersaFlash Videos > VersaPak Cartridge Range
VersaFlash™ Videos
VersaPak Cartridge Range
Video displays the wide range of VersaPak® cartridge dimensions that can be used with the VersaFlash® system, which corresponds to a sample handling capability from mg to kg.
Analytical Seminars
Sign up to attend a live seminar!
| VersaFlash Products | Request VersaFlash CD |
| Video Feedback |
video with Brazil Portuguese Vídeo com tradução para Português
The most unique feature of the VersaFlash system is its broad range of sample-handling capabilities. The VersaFlash system allows you to work with 110 by 300 mm 1.35 kilogram cartridges to purify over 100-gram samples. Then within seconds you can scale down and efficiently work with 23 by 53 mm cartridges to purify single and sub-gram quantity samples. Unlike other systems that either restrict the range of cartridges and samples that you can work with, or require manipulation of compression barrels and other cumbersome hardware, scale up with the VersaFlash system is accomplished through simple mechanical adjustments.