Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Videos & Web Seminars > VersaFlash Videos > VersaVac Sample Loading Station
VersaFlash™ Videos
VersaVac Sample Loading Station
This video shows how the VersaVac® vacuum manifold station is used to draw a sample onto a flash cartridge, an optional sample loading procedure.
Analytical Seminars
Sign up to attend a live seminar!
| VersaFlash Products | Request VersaFlash CD |
| Video Feedback |
video with Brazil Portuguese Vídeo com tradução para Português
The VersaFlash system is also available with the VersaVac sample loading station, which is a vacuum manifold that allows you to draw the sample onto a cartridge, using vacuum. With this unit, a syringe barrel can be used as a sample reservoir or an SPE tube as a pre-purification step. Vacuum is applied to introduce the sample onto the purification cartridge. The vacuum is then released, and the cartridge is ready to load into the VersaFlash station. Additionally, the VersaVac can be used to empty used cartridges from solvent for simpler waste disposal. The VersaVac is compatible with the full range of cartridge sizes.