EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Chemically-induced apoptosis: p21 and p53 as determinants of enterotoxin activity.
PMID 10022227
Abstract
The relationship between toxin-induced apoptosis and longer-term (> 72 h) intestinal toxicity was investigated in vivo using p53 wild type (+/+) and 'knockout' (-/-) mice. The enterotoxic antimetabolite 5-fluorouracil (5-FU) induced acute p53-dependent apoptosis in the crypts of both small intestine and midcolon. Although the amount of apoptosis was the same order of magnitude at its peak (24 h) at both 40 and 400 mg/kg 5-FU, only 400 mg/kg 5-FU brought about changes in the integrity of the gut after 96 h. These were characterised by the loss of epithelial cells from crypts and villi. Only after 400 mg/kg 5-FU were mitotic index and DNA synthesis significantly suppressed in both small intestinal and midcolonic crypts. This correlated with a prolonged, p53-dependent expression of p21(waf-1/cip1). In p53 null (-/-) mice significant reductions in 5-FU-induced apoptosis and relief from the inhibition of cell cycle progression permitted retention of crypt integrity after 5-FU. Thus, quantitative measures of acute apoptosis in vivo did not accurately predict subsequent pathological changes in the gut. Rather, p53-dependent inhibition of cell cycle progression together with cell loss by apoptosis caused a loss of crypt integrity. Importantly, the tissue toxicity of 5-fluorouracil was genetically determined at a locus (p53) separate from that directly associated with toxin action. The selectivity of toxin action is therefore also determined by events 'downstream' of those associated with the direct mode of action of the toxin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6627
5-Fluorouracil, ≥99% (HPLC), powder
C4H3FN2O2