EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Metabolism of the new liposomal anticancer drug N4-octadecyl-1-beta-D-arabinofuranosylcytosine in mice.
PMID 10064564
Abstract
Metabolism and excretion of the new antitumor drug N4-octadecyl-1-beta-D-arabinofuranosylcytosine (NOAC) was investigated in mice. Mice were injected i.v. with tritium-labeled liposomal NOAC (4 micromol/mouse). Analysis of HPLC-purified extracts of liver homogenates by liquid chromatography coupled with mass spectrometry revealed only the presence of unmetabolized drug. To study the excretion of the administered drug, mice were injected with tritium-labeled liposomal NOAC or as comparison with 1-beta-D-arabinofuranosylcytosine (ara-C; 4 micromol/mouse) and housed up to 48 h in metabolic cages. Urine and feces were collected at different time points and the kinetics of excreted radioactivity were determined. After 48 h, 39% of the injected [5-3H]NOAC radioactivity was excreted in urine and 16% in feces, whereas ara-C radioactivity was only found in urine with 48% of the injected dose. Feces extracts and urine were purified by HPLC and radioactive fractions were further analyzed by liquid chromatography coupled with mass spectrometry. The radioactivity of feces extracts of NOAC-treated mice was composed of unmetabolized NOAC, hydroxylated NOAC (NOAC + OH), its sulfated derivative (NOAC + OSO3H), and unidentified metabolites, whereas in urine, the hydrophilic molecules ara-C and ara-U were found. During the period of 48 h only 2% of the injected NOAC was eliminated in its unmetabolized form, whereas 25% was identified as main metabolite ara-C. Urine collected during 48 h in ara-C-treated mice contained 33% of the injected dose as unmetabolized drug and 13% as the main metabolite ara-U. Thus, NOAC is metabolized by two major pathways, one leading to the hydrophilic metabolites ara-C and ara-U and the other to hydroxylated and sulfated NOAC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O5508
N4-Octadecylcytosine β-D-arabinofuranoside
C27H49N3O5