EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Detailed marker chromosome analysis in cell line U-BLC1, established from transitional-cell carcinoma of the bladder.
PMID 10074925
Abstract
A permanent cell line, U-BLC1, was established from a primary transitional-cell carcinoma, TCC, of the urinary bladder. Karyotype analysis showed the line to be highly aberrant, with a near-triploid chromosome number of 68 to 73. Comparative genomic hybridization revealed some distinct differences between the primary tumor and the established cell line. Karyotype analysis showed 3 marker chromosomes with homogeneously staining regions, HSRs, in the cell line. The HSRs were isolated by microdissection and the microdissection probes were hybridized to normal metaphase chromosomes. The HSRs contain sequences known to be frequently involved in amplification in transitional-cell carcinoma of the bladder, 6p22, 7p11-p12, 9p23-pter, and one region not yet reported to be amplified in primary TCC of the bladder, 1p31-p32. A candidate-gene approach showed that in the region 7p11-p12 the EGFR locus is amplified and highly expressed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06013102
U-BLC1