EMAIL THIS PAGE TO A FRIEND
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology
The effects of benzodiazepine (triazolam), cyclopyrrolone (zopiclone) and imidazopyridine (zolpidem) hypnotics on the frequency of hippocampal theta activity and sleep structure in rats.
PMID 10082225
Abstract
In order to investigate the relative efficacy and safety of zopiclone and zolpidem, we compared the effects of higher doses of zopiclone and zolpidem on the frequency of hippocampal theta activity and sleep structure with that of triazolam. Rats were divided into triazolam treatment group (1 mg/kg, 5 mg/kg), zopiclone treatment group (20 mg/kg, 100 mg/kg) and zolpidem treatment group (20 mg/kg, 100 mg/kg). Rats were injected intraperitoneally with these drugs or their vehicle. Polygraphic sleep recording and visual frequency analysis of the hippocampal EEG activity in REM sleep were carried out for 6 h after each injection. Zolpidem, unlike triazolam and zopiclone, had a much milder reducing-effect on the frequency of hippocampal theta activity and suppressing-effect on REM sleep. These results suggest that zolpidem may prove to be a safer hypnotic drug which has fewer or milder side effects than are benzodiazepine and cyclopyrrolone hypnotics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T9772
Triazolam
C17H12Cl2N4
Z4900
Zopiclone
C17H17ClN6O3