EMAIL THIS PAGE TO A FRIEND
Operative dentistry
Hardening of new resin cements cured through a ceramic inlay.
PMID 10337297
Abstract
This study investigated the degree of hardening achieved through self-curing only and through dual-curing a group of eight new resin-based cements. In addition, the effect of ceramic inlay thickness on cement hardness was determined. Disk specimens measuring 6 mm in diameter and 2.5 mm thick were prepared from eight cements: Adherence, Choice, Duolink, Enforce, Lute-It, Nexus, Resinomer, and Variolink. Eight specimens were prepared from each material; half were self-cured, while the remainder were dual-cured. Knoop hardness measurements were then made at 1-hour, 1-day, and 1-week intervals. In addition 12 specimens of the same dimensions were prepared from each cement and were dual-cured through ceramic spacers of varying thickness (1-6 mm). Hardness measurements were made as above. ANOVA showed significant differences in hardness of self-cured versus dual-cured specimens for all cements (P < 0.0001). Significant differences were also found in the hardness of specimens dual-cured through ceramic spacers 2-3 mm in thickness or more compared with those that were dual-cured without spacer. It is concluded that for some materials self-curing alone was not adequate to achieve sufficient hardening; cement hardness was significantly reduced when ceramic inlay thickness was 2-3 mm or more.