EMAIL THIS PAGE TO A FRIEND
Therapeutic drug monitoring
Determination of mitotane (o,p-DDD) and its metabolites o,p-DDA and o,p-DDE in plasma by high-performance liquid chromatography.
PMID 10365653
Abstract
The adrenolytic agent mitotane (o,p'-DDD or 1,1-(o,p'-dichlorodiphenyl)-2,2-dichoroethane) is believed to be metabolically activated, resulting in the generation of reactive intermediates or the formation of o,p'-DDA. A new high-performance liquid chromatographic (HPLC) method for measurement of mitotane and two of its metabolites, o,p'-DDA (1,1-(o,p'-dichlorodiphenyl) acetic acid) and o,p'-DDE (1,1-(o,p'-dichlorodiphenyl)-2,2 dichloroethene), in plasma has been developed. Sample preparation involves precipitation of plasma proteins with acetone before chromatographic separation under isocratic conditions on a silica-based diphenyl column. Mitotane and metabolites are quantified by ultraviolet detection at 218 nm. Recoveries for the three compounds range between 99% and 110%, with between-day and within-day coefficients of variation <6% within the therapeutic range. Limits of detection are 0.5 microM and the assay is linear up to at least 800 microM for o,p'-DDA and 100 microM for mitotane and o,p'-DDE. The method has been evaluated using samples from three patients on mitotane therapy, revealing o,p'-DDA levels in plasma 3 to 10 times higher than the levels of the parent compound.