EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
Naloxone blocks endomorphin-1 but not endomorphin-2 induced inhibition of tachykinergic contractions of guinea-pig isolated bronchus.
PMID 10401549
Abstract
The recently identified endogenous agonists on the mu-opioid-receptor (mu OR), endomorphin-1 (EM-1) and endomorphin-2 (EM-2), induce a concentration dependent inhibition of electrical field stimulation (EFS)-induced tachykinin-mediated contractions of the guinea-pig bronchus (ED50s < 10 nM for both compounds). Surprisingly, only endomorphin-1 effects could be blocked by naloxone (10 microM), whereas endomorphin-2 effects were not affected by specific antagonists for the mu-, kappa-, and delta-opioid-receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7758
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder
C19H21NO4 · HCl · 2H2O