EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Daphnetin, one of coumarin derivatives, is a protein kinase inhibitor.
PMID 10403826
Abstract
Protein kinases play key roles in the control of cell proliferation, differentiation and metabolism. In this work, we studied the effect of coumarin and its derivatives, including daphnetin, esculin, 2-OH-coumarin, 4-OH-coumarin and 7-OH-coumarin, on the activity of protein kinases. It was found that, in these compounds, only daphnetin was a protein kinase inhibitor. This compound inhibited tyrosine-specific protein kinase, EGF receptor (IC(50) = 7.67 microM), and serine/threonine-specific protein kinases, including cAMP-dependent protein kinase (PKA) (IC(50) = 9.33 microM) and protein kinase C (PKC) (IC(50) = 25.01 microM) in vitro. The inhibition of EGF receptor tyrosine kinase by daphnetin was competitive to ATP and non-competitive to the peptide substrate. The inhibition of EGF-induced tyrosine phosphorylation of EGF receptor by daphnetin was not observed in human hepatocellular carcinoma HepG2 cells. The structural comparison of daphnetin with coumarin and other coumarin derivatives suggests that the hydroxylation at C8 may be required for daphnetin acting as a protein kinase inhibitor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5564
7,8-Dihydroxycoumarin, ≥97%, powder
C9H6O4