EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
The MUC3 gene encodes a transmembrane mucin and is alternatively spliced.
PMID 10405327
Abstract
Epithelial mucins are a family of secreted and cell surface glycoproteins expressed by epithelial tissues and implicated in epithelial cell protection, adhesion modulation and signaling. The gene encoding human MUC3 (hMUC3), localised to chromosome 7q22, is most highly expressed in the small intestine. It has previously been reported to be a non-transmembrane mucin with minimal homology to its suggested orthologues from rat (rMuc3) and mouse (mMuc3). RT-PCR was performed to investigate the carboxyl terminus of the published sequence of hMUC3 from normal colon and small intestine tissues and also from a series of 10 colorectal cancer cell lines. Two distinct PCR products were identified. In contrast to the previously published hMUC3 sequence, which terminates shortly after a single cysteine-rich EGF-like domain, conceptual protein translation of the dominant and largest PCR product identified two extracellular cysteine-rich EGF-like domains separated by an N-glycosylation-rich domain and a potential coiled-coil region, followed by a putative transmembrane region and a 75 amino acid cytoplasmic tail. The smaller of the two PCR products was found to be an alternative splice variant of MUC3 including the first EGF-like domain but lacking part of the second EGF-like domain and the transmembrane region. Nine out of 10 colorectal cancer cell lines were found to express MUC3. Interestingly, one of the cell lines, LoVo, expressed predominantly the alternative splice form lacking a transmembrane domain. Structural homology of the new protein sequence of hMUC3 with rMuc3 and mMuc3 indicates it is closely related to the rodent proteins and is likely to be involved in ligand-binding and intracellular signaling. The new finding that MUC3 encodes a transmembrane molecule presents a new paradigm for the structure of this mucin and the manner in which it may function.