EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Anticonvulsant-resistant seizures following pyridostigmine bromide (PB) and N,N-diethyl-m-toluamide (DEET).
PMID 10416276
Abstract
An acute toxic interaction has been described, in which sublethal doses of pyridostigmine bromide (PB) and the insect repellent N,N-diethyl-m-toluamide (DEET), when administered concomitantly, resulted in seizures and lethality. To investigate the possible relationships between seizures and lethality and the role of the cholinergic system in this interaction, PB (5 mg/kg), DEET (200 mg/kg) or PB (3 mg/kg) + DEET (200 mg/kg) were administered i.p. to male ICR mice, alone or following i.p. pretreatment, with one of several anticonvulsant agents: diazepam, 10 mg/kg; fosphenytoin, 40 mg/kg; phenobarbital, 45 mg/kg; or dextrophan, 25 mg/kg), or the anticholinergic agents, atropine (5 mg/kg), atropine methyl nitrate (2.7 mg/kg), or mecamylamine (2.5 mg/kg). The anticonvulsants selected for this study act through different mechanisms to reduce seizures. None of the anticonvulsants was able to reduce the incidence of seizures following treatment with PB, DEET or PB + DEET. Only diazepam delayed the onset of seizures. Fosphenytoin or diazepam significantly prolonged the time to lethality following PB, but only fosphenytoin reduced the incidence of PB-induced lethality. Diazepam or phenobarbital significantly prolonged the time to lethality following PB + DEET. Both atropine and atropine methyl nitrate protected against PB and PB + DEET-induced lethality and PB-induced seizures. Neither agent blocked seizures resulting from DEET or PB + DEET. Mecamylamine reduced seizures and lethality in PB-treated mice, but not in mice treated with DEET or PB + DEET. The results indicate that seizure activity is not a causative factor in the toxic interaction between PB and DEET. Furthermore, PB, DEET and PB + DEET induce seizures that are resistant to standard anticonvulsants, and each appears to operate through different mechanisms to produce seizures. Peripheral muscarinic receptors may play a specific role in lethality caused by PB + DEET.