EMAIL THIS PAGE TO A FRIEND
Journal of cardiovascular pharmacology
Impaired renal vascular response to a D1-like receptor agonist but not to an ACE inhibitor in conscious spontaneously hypertensive rats.
PMID 10445669
Abstract
The natriuretic response to a dopamine 1-like receptor agonist is blunted in spontaneously hypertensive rats (SHRs). Whether the renal vasodilator response to D1-like receptor stimulation in SHRs is defective also is unclear. To determine whether the renal hemodynamic response to a D1-like receptor is impaired in SHR, we examined the effect of a continuous infusion of the D1-like receptor agonist fenoldopam (2 microg/kg/min) on systemic and renal hemodynamics in conscious SHRs and Wistar-Kyoto (WKY) rats. As an active control, we used an equivalent antihypertensive dosage of captopril (10 mg/kg). Fenoldopam significantly increased effective renal plasma flow (ERPF) in WKY rats (+22 +/- 5%; p < 0.01), whereas this response was absent in SHRs (+7 +/- 3%; NS). Mean arterial pressure (MAP) was significantly reduced in SHRs (-11 +/- 2%; p < 0.001), demonstrating a systemic vasodilator response to fenoldopam in SHRs. The reduction in renal vascular resistance (RVR) was more pronounced in WKY rats (-24 +/- 2%) than in SHRs (-13 +/- 4%; p < 0.05). Captopril significantly increased ERPF in SHRs (+16 +/- 3%; p < 0.001), demonstrating a preserved renal vasodilatory capacity in SHRs. The blunting of the renal vasodilatory response to fenoldopam in SHRs is present during a high as well as a low sodium intake. In conscious SHRs, the renal vasodilatory response to a D1-like receptor agonist is impaired, whereas the blood pressure response is more pronounced. The preserved renal vasodilatory response to captopril indicates that the defective vasodilatory response in SHRs is functional rather than due to altered structural properties of the renal vascular bed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6800
Fenoldopam monohydrobromide, ≥98%
C16H16ClNO3 · HBr